Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81332252-81332517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT12
Entrez Gene ID
134526
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
CACH-1, Cach, STARD15, THEAL
UniGene
Hs.591756
Gene Chromosome Location
Chr.5: 81309045-81394174 on Build GRCh38
Transcripts
NM_130767.2 XM_006714531.2 XM_006714532.3 XM_017009045.1 XM_017009046.1 XM_017009047.1 XM_017009048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618363 c.1370G>A p.R457Q Chr.5: 81332498-81332498 on Build GRCh38
3618362 c.1380C>T p.P460P Chr.5: 81332488-81332488 on Build GRCh38
4437059 c.1367C>T p.S456L Chr.5: 81332501-81332501 on Build GRCh38
7093920 c.1362C>T p.L454L Chr.5: 81332506-81332506 on Build GRCh38
5466587 c.1367C>A p.S456* Chr.5: 81332501-81332501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983717072 Chr.5: 81332410-81332410 on Build GRCh38
rs536220210 Chr.5: 81332372-81332372 on Build GRCh38
rs748438264 Chr.5: 81332451-81332451 on Build GRCh38
rs147089882 Chr.5: 81332483-81332483 on Build GRCh38
rs150336466 Chr.5: 81332408-81332408 on Build GRCh38
rs755384770 Chr.5: 81332443-81332443 on Build GRCh38
rs779239549 Chr.5: 81332448-81332448 on Build GRCh38
rs974527357 Chr.5: 81332289-81332289 on Build GRCh38
rs73134848 Chr.5: 81332385-81332385 on Build GRCh38
rs867130905 Chr.5: 81332416-81332416 on Build GRCh38
rs1034521600 Chr.5: 81332366-81332366 on Build GRCh38
rs372250212 Chr.5: 81332432-81332432 on Build GRCh38
rs758591450 Chr.5: 81332465-81332465 on Build GRCh38
rs557892282 Chr.5: 81332484-81332484 on Build GRCh38
rs141598728 Chr.5: 81332478-81332478 on Build GRCh38
rs950848094 Chr.5: 81332401-81332401 on Build GRCh38
rs73134843 Chr.5: 81332298-81332298 on Build GRCh38
rs555829689 Chr.5: 81332383-81332383 on Build GRCh38

View Full Product Details