Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8764685-8764918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABAT
Entrez Gene ID
18
Gene Name
4-aminobutyrate aminotransferase
Gene Aliases
GABA-AT, GABAT, NPD009
UniGene
Hs.336768
Gene Chromosome Location
Chr.16: 8674587-8784575 on Build GRCh38
Transcripts
NM_000663.4 NM_001127448.1 NM_020686.5 XM_011522400.2 XM_011522401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
98100 c.511G>A p.A171T Chr.16: 8764801-8764801 on Build GRCh38
974701 c.539G>A p.R180Q Chr.16: 8764829-8764829 on Build GRCh38
974700 c.465G>A p.M155I Chr.16: 8764755-8764755 on Build GRCh38
5055166 c.490G>A p.G164S Chr.16: 8764780-8764780 on Build GRCh38
6304280 c.510C>T p.N170N Chr.16: 8764800-8764800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755726683 Chr.16: 8764866-8764870 on Build GRCh38
rs757056947 Chr.16: 8764793-8764793 on Build GRCh38
rs148949594 Chr.16: 8764843-8764844 on Build GRCh38
rs746589996 Chr.16: 8764828-8764828 on Build GRCh38
rs910548156 Chr.16: 8764892-8764892 on Build GRCh38
rs766966035 Chr.16: 8764846-8764846 on Build GRCh38
rs762214989 Chr.16: 8764838-8764838 on Build GRCh38
rs200627621 Chr.16: 8764779-8764779 on Build GRCh38
rs147628671 Chr.16: 8764787-8764787 on Build GRCh38
rs759615487 Chr.16: 8764853-8764853 on Build GRCh38
rs750283443 Chr.16: 8764872-8764872 on Build GRCh38
rs35745596 Chr.16: 8764865-8764866 on Build GRCh38
rs749138260 Chr.16: 8764717-8764717 on Build GRCh38
rs777322178 Chr.16: 8764866-8764875 on Build GRCh38
rs745824946 Chr.16: 8764811-8764811 on Build GRCh38
rs779411867 Chr.16: 8764821-8764821 on Build GRCh38
rs762337644 Chr.16: 8764761-8764761 on Build GRCh38
rs780108604 Chr.16: 8764879-8764879 on Build GRCh38
rs771506614 Chr.16: 8764727-8764727 on Build GRCh38
rs755579396 Chr.16: 8764844-8764849 on Build GRCh38
rs903118661 Chr.16: 8764801-8764801 on Build GRCh38
rs750449172 Chr.16: 8764844-8764847 on Build GRCh38
rs2302608 Chr.16: 8764723-8764723 on Build GRCh38
rs141981377 Chr.16: 8764800-8764800 on Build GRCh38
rs776431473 Chr.16: 8764829-8764829 on Build GRCh38
rs375677145 Chr.16: 8764719-8764719 on Build GRCh38
rs750911984 Chr.16: 8764774-8764774 on Build GRCh38
rs751707478 Chr.16: 8764782-8764782 on Build GRCh38
rs767668242 Chr.16: 8764860-8764860 on Build GRCh38
rs1056510848 Chr.16: 8764736-8764736 on Build GRCh38
rs572662581 Chr.16: 8764860-8764861 on Build GRCh38
rs896483658 Chr.16: 8764746-8764746 on Build GRCh38
rs200644195 Chr.16: 8764718-8764718 on Build GRCh38
rs757007502 Chr.16: 8764868-8764868 on Build GRCh38
rs890043703 Chr.16: 8764878-8764878 on Build GRCh38
rs369503600 Chr.16: 8764713-8764713 on Build GRCh38
rs772087969 Chr.16: 8764819-8764819 on Build GRCh38
rs765389727 Chr.16: 8764842-8764843 on Build GRCh38
rs892963068 Chr.16: 8764752-8764752 on Build GRCh38
rs112675542 Chr.16: 8764844-8764845 on Build GRCh38
rs753562528 Chr.16: 8764791-8764791 on Build GRCh38
rs374659543 Chr.16: 8764733-8764733 on Build GRCh38
rs777126176 Chr.16: 8764751-8764751 on Build GRCh38
rs770310062 Chr.16: 8764844-8764844 on Build GRCh38
rs527558391 Chr.16: 8764891-8764891 on Build GRCh38
rs58334184 Chr.16: 8764870-8764870 on Build GRCh38
rs755732557 Chr.16: 8764728-8764728 on Build GRCh38
rs751757406 Chr.16: 8764708-8764708 on Build GRCh38
rs763045532 Chr.16: 8764776-8764776 on Build GRCh38
rs755177048 Chr.16: 8764783-8764783 on Build GRCh38
rs773662763 Chr.16: 8764845-8764845 on Build GRCh38
rs758257577 Chr.16: 8764877-8764877 on Build GRCh38
rs774289583 Chr.16: 8764847-8764847 on Build GRCh38
rs752938327 Chr.16: 8764861-8764861 on Build GRCh38
rs1012666327 Chr.16: 8764844-8764853 on Build GRCh38

View Full Product Details