Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34101563-34101820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC7
Entrez Gene ID
56851
Gene Name
ER membrane protein complex subunit 7
Gene Aliases
C11orf3, C15orf24, HT022, ORF1-FL1
UniGene
Hs.160565
Gene Chromosome Location
Chr.15: 34084022-34101852 on Build GRCh38
Transcripts
NM_020154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092065 c.53C>T p.S18L Chr.15: 34101787-34101787 on Build GRCh38
7114740 c.66G>A p.Q22Q Chr.15: 34101774-34101774 on Build GRCh38
1743250 c.133T>C p.F45L Chr.15: 34101707-34101707 on Build GRCh38
6653636 c.128A>G p.D43G Chr.15: 34101712-34101712 on Build GRCh38
4054102 c.85G>A p.A29T Chr.15: 34101755-34101755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764550728 Chr.15: 34101654-34101654 on Build GRCh38
rs751035105 Chr.15: 34101675-34101675 on Build GRCh38
rs550063207 Chr.15: 34101760-34101760 on Build GRCh38
rs550543116 Chr.15: 34101708-34101708 on Build GRCh38
rs866434424 Chr.15: 34101739-34101739 on Build GRCh38
rs759705686 Chr.15: 34101716-34101716 on Build GRCh38
rs777828275 Chr.15: 34101789-34101790 on Build GRCh38
rs571422272 Chr.15: 34101586-34101586 on Build GRCh38
rs757683222 Chr.15: 34101667-34101667 on Build GRCh38
rs527261396 Chr.15: 34101756-34101756 on Build GRCh38
rs77698400 Chr.15: 34101726-34101726 on Build GRCh38
rs764975902 Chr.15: 34101751-34101751 on Build GRCh38
rs765675291 Chr.15: 34101772-34101772 on Build GRCh38
rs1048796154 Chr.15: 34101758-34101758 on Build GRCh38
rs761182498 Chr.15: 34101644-34101644 on Build GRCh38
rs754425342 Chr.15: 34101676-34101676 on Build GRCh38
rs536183649 Chr.15: 34101769-34101769 on Build GRCh38
rs753053065 Chr.15: 34101673-34101673 on Build GRCh38
rs549259556 Chr.15: 34101678-34101678 on Build GRCh38
rs750915064 Chr.15: 34101782-34101782 on Build GRCh38
rs948063561 Chr.15: 34101591-34101591 on Build GRCh38
rs868419006 Chr.15: 34101721-34101721 on Build GRCh38
rs777404352 Chr.15: 34101706-34101706 on Build GRCh38
rs767861151 Chr.15: 34101634-34101634 on Build GRCh38
rs552729650 Chr.15: 34101585-34101585 on Build GRCh38
rs772268121 Chr.15: 34101718-34101718 on Build GRCh38
rs770350127 Chr.15: 34101713-34101713 on Build GRCh38
rs770809700 Chr.15: 34101712-34101712 on Build GRCh38
rs779436277 Chr.15: 34101669-34101669 on Build GRCh38
rs769614065 Chr.15: 34101702-34101702 on Build GRCh38
rs747791884 Chr.15: 34101688-34101688 on Build GRCh38
rs767998130 Chr.15: 34101626-34101628 on Build GRCh38
rs759652763 Chr.15: 34101608-34101608 on Build GRCh38
rs754411124 Chr.15: 34101666-34101666 on Build GRCh38
rs764569500 Chr.15: 34101741-34101741 on Build GRCh38
rs554533846 Chr.15: 34101795-34101795 on Build GRCh38
rs756436394 Chr.15: 34101790-34101792 on Build GRCh38
rs780879341 Chr.15: 34101686-34101686 on Build GRCh38
rs532357128 Chr.15: 34101638-34101638 on Build GRCh38

View Full Product Details