Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54372243-54372504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
294189 c.878C>A p.P293H Chr.18: 54372500-54372500 on Build GRCh38
3668135 c.867A>G p.K289K Chr.18: 54372489-54372489 on Build GRCh38
988989 c.876T>C p.G292G Chr.18: 54372498-54372498 on Build GRCh38
5576463 c.860G>A p.G287E Chr.18: 54372482-54372482 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371851488 Chr.18: 54372468-54372468 on Build GRCh38
rs1017192799 Chr.18: 54372382-54372382 on Build GRCh38
rs777254306 Chr.18: 54372477-54372477 on Build GRCh38
rs969423500 Chr.18: 54372475-54372475 on Build GRCh38
rs759562914 Chr.18: 54372415-54372415 on Build GRCh38
rs767669784 Chr.18: 54372441-54372441 on Build GRCh38
rs764164918 Chr.18: 54372313-54372313 on Build GRCh38
rs555759574 Chr.18: 54372299-54372299 on Build GRCh38
rs754275952 Chr.18: 54372449-54372449 on Build GRCh38
rs750686713 Chr.18: 54372443-54372443 on Build GRCh38
rs766457467 Chr.18: 54372448-54372448 on Build GRCh38
rs761627130 Chr.18: 54372440-54372441 on Build GRCh38
rs566361711 Chr.18: 54372304-54372304 on Build GRCh38
rs959322500 Chr.18: 54372321-54372321 on Build GRCh38
rs756133590 Chr.18: 54372446-54372446 on Build GRCh38
rs755546192 Chr.18: 54372450-54372450 on Build GRCh38
rs368958988 Chr.18: 54372431-54372431 on Build GRCh38
rs987507471 Chr.18: 54372479-54372479 on Build GRCh38
rs755143134 Chr.18: 54372357-54372357 on Build GRCh38
rs554555629 Chr.18: 54372293-54372293 on Build GRCh38
rs1034891849 Chr.18: 54372311-54372311 on Build GRCh38
rs748510037 Chr.18: 54372470-54372470 on Build GRCh38
rs775218567 Chr.18: 54372428-54372428 on Build GRCh38
rs558297685 Chr.18: 54372473-54372473 on Build GRCh38
rs765061718 Chr.18: 54372416-54372416 on Build GRCh38
rs1031205281 Chr.18: 54372398-54372398 on Build GRCh38
rs963471097 Chr.18: 54372383-54372383 on Build GRCh38
rs191353715 Chr.18: 54372352-54372352 on Build GRCh38
rs34754885 Chr.18: 54372343-54372344 on Build GRCh38
rs919229911 Chr.18: 54372460-54372460 on Build GRCh38
rs999267063 Chr.18: 54372355-54372358 on Build GRCh38

View Full Product Details