Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19385556-19385789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZB
Entrez Gene ID
832
Gene Name
capping actin protein of muscle Z-line beta subunit
Gene Aliases
CAPB, CAPPB, CAPZ
UniGene
Hs.432760
Gene Chromosome Location
Chr.1: 19338773-19485641 on Build GRCh38
Transcripts
NM_001206540.2 NM_001206541.2 NM_001282162.1 NM_001313932.1 NM_004930.4 XM_006710938.3 XM_011542228.2 XM_011542229.2 XM_011542230.2 XM_017002428.1 XM_017002429.1 XM_017002430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4771624 c.209A>C p.D70A Chr.1: 19385589-19385589 on Build GRCh38
4771623 c.131A>C p.D44A Chr.1: 19385589-19385589 on Build GRCh38
2157668 c.134_135insC p.Q45fs*19 Chr.1: 19385585-19385586 on Build GRCh38
4771622 c.131A>C p.D44A Chr.1: 19385589-19385589 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756604515 Chr.1: 19385757-19385757 on Build GRCh38
rs10917439 Chr.1: 19385751-19385751 on Build GRCh38
rs571826508 Chr.1: 19385594-19385594 on Build GRCh38
rs781519276 Chr.1: 19385699-19385699 on Build GRCh38
rs185311361 Chr.1: 19385650-19385650 on Build GRCh38
rs775479259 Chr.1: 19385729-19385729 on Build GRCh38
rs1055904827 Chr.1: 19385682-19385682 on Build GRCh38
rs755330132 Chr.1: 19385754-19385754 on Build GRCh38
rs757283042 Chr.1: 19385630-19385630 on Build GRCh38
rs764104988 Chr.1: 19385615-19385615 on Build GRCh38
rs756629065 Chr.1: 19385651-19385651 on Build GRCh38
rs773844823 Chr.1: 19385708-19385708 on Build GRCh38
rs745538853 Chr.1: 19385720-19385720 on Build GRCh38
rs74941542 Chr.1: 19385721-19385721 on Build GRCh38
rs917151266 Chr.1: 19385759-19385759 on Build GRCh38
rs371154610 Chr.1: 19385746-19385746 on Build GRCh38
rs924448825 Chr.1: 19385747-19385747 on Build GRCh38
rs755525971 Chr.1: 19385681-19385681 on Build GRCh38
rs532876346 Chr.1: 19385620-19385620 on Build GRCh38
rs775385399 Chr.1: 19385589-19385589 on Build GRCh38
rs764197063 Chr.1: 19385732-19385732 on Build GRCh38
rs115110021 Chr.1: 19385756-19385756 on Build GRCh38
rs373710207 Chr.1: 19385679-19385679 on Build GRCh38
rs750472553 Chr.1: 19385745-19385745 on Build GRCh38
rs776416481 Chr.1: 19385734-19385734 on Build GRCh38
rs11555671 Chr.1: 19385592-19385592 on Build GRCh38
rs765232350 Chr.1: 19385742-19385742 on Build GRCh38
rs761749853 Chr.1: 19385739-19385739 on Build GRCh38
rs748889242 Chr.1: 19385701-19385701 on Build GRCh38
rs375680442 Chr.1: 19385581-19385581 on Build GRCh38
rs189967158 Chr.1: 19385674-19385674 on Build GRCh38
rs766564661 Chr.1: 19385749-19385749 on Build GRCh38
rs770362321 Chr.1: 19385707-19385707 on Build GRCh38
rs898737339 Chr.1: 19385713-19385713 on Build GRCh38
rs761650418 Chr.1: 19385656-19385657 on Build GRCh38
rs758545440 Chr.1: 19385670-19385670 on Build GRCh38
rs200839344 Chr.1: 19385633-19385633 on Build GRCh38
rs993754632 Chr.1: 19385625-19385625 on Build GRCh38
rs765551537 Chr.1: 19385635-19385635 on Build GRCh38

View Full Product Details