Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48847983-48848210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO2
Entrez Gene ID
10241
Gene Name
calcium binding and coiled-coil domain 2
Gene Aliases
NDP52
UniGene
Hs.514920
Gene Chromosome Location
Chr.17: 48830988-48865245 on Build GRCh38
Transcripts
NM_001261390.1 NM_001261391.1 NM_001261393.1 NM_001261395.1 NM_005831.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6619016 c.217A>G p.M73V Chr.17: 48848100-48848100 on Build GRCh38
5508891 c.254C>T p.S85L Chr.17: 48848137-48848137 on Build GRCh38
3518806 c.198C>T p.T66T Chr.17: 48848081-48848081 on Build GRCh38
5958737 c.240C>G p.D80E Chr.17: 48848123-48848123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767270527 Chr.17: 48848172-48848172 on Build GRCh38
rs528884391 Chr.17: 48848151-48848151 on Build GRCh38
rs778232629 Chr.17: 48848091-48848091 on Build GRCh38
rs751043237 Chr.17: 48848106-48848106 on Build GRCh38
rs756963067 Chr.17: 48848048-48848048 on Build GRCh38
rs186495533 Chr.17: 48848061-48848061 on Build GRCh38
rs771231561 Chr.17: 48848094-48848094 on Build GRCh38
rs540353221 Chr.17: 48848131-48848131 on Build GRCh38
rs926668850 Chr.17: 48848137-48848137 on Build GRCh38
rs374335998 Chr.17: 48848093-48848093 on Build GRCh38
rs775486120 Chr.17: 48848125-48848125 on Build GRCh38
rs201620728 Chr.17: 48848082-48848082 on Build GRCh38
rs947108155 Chr.17: 48848158-48848158 on Build GRCh38
rs939177837 Chr.17: 48848159-48848159 on Build GRCh38
rs755774644 Chr.17: 48848185-48848185 on Build GRCh38
rs763435753 Chr.17: 48848057-48848057 on Build GRCh38
rs779648908 Chr.17: 48848080-48848080 on Build GRCh38
rs1031636300 Chr.17: 48848127-48848127 on Build GRCh38
rs375935534 Chr.17: 48848083-48848083 on Build GRCh38
rs990054297 Chr.17: 48848145-48848147 on Build GRCh38
rs755684681 Chr.17: 48848065-48848065 on Build GRCh38
rs377145641 Chr.17: 48848116-48848116 on Build GRCh38
rs191324287 Chr.17: 48848181-48848181 on Build GRCh38
rs767090472 Chr.17: 48848039-48848039 on Build GRCh38
rs751408711 Chr.17: 48848147-48848147 on Build GRCh38
rs560158466 Chr.17: 48848138-48848138 on Build GRCh38
rs759678667 Chr.17: 48848110-48848110 on Build GRCh38
rs367731456 Chr.17: 48848102-48848102 on Build GRCh38
rs750015045 Chr.17: 48848064-48848064 on Build GRCh38

View Full Product Details