Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186852852-186853125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOQ
Entrez Gene ID
9370
Gene Name
adiponectin, C1Q and collagen domain containing
Gene Aliases
ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28
UniGene
Hs.80485
Gene Chromosome Location
Chr.3: 186842674-186858463 on Build GRCh38
Transcripts
NM_001177800.1 NM_004797.3 NR_046662.1

Gene Symbol
ADIPOQ-AS1
Entrez Gene ID
100874095
Gene Name
ADIPOQ antisense RNA 1
Gene Aliases
-
UniGene
Hs.638968
Gene Chromosome Location
Chr.3: 186851886-186856123 on Build GRCh38
Transcripts
NM_001177800.1 NM_004797.3 NR_046662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7221439 c.45T>G p.G15G Chr.3: 186853103-186853103 on Build GRCh38
3591049 c.63G>A p.T21T Chr.3: 186853121-186853121 on Build GRCh38
6097378 c.44G>T p.G15V Chr.3: 186853102-186853102 on Build GRCh38
5708644 c.62C>T p.T21M Chr.3: 186853120-186853120 on Build GRCh38
5615250 c.67C>G p.Q23E Chr.3: 186853125-186853125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138678413 Chr.3: 186853100-186853100 on Build GRCh38
rs566914377 Chr.3: 186852967-186852967 on Build GRCh38
rs1018755353 Chr.3: 186852890-186852890 on Build GRCh38
rs964238268 Chr.3: 186853059-186853059 on Build GRCh38
rs114155159 Chr.3: 186853084-186853084 on Build GRCh38
rs576839771 Chr.3: 186852999-186852999 on Build GRCh38
rs959782510 Chr.3: 186852992-186852992 on Build GRCh38
rs752180988 Chr.3: 186853098-186853098 on Build GRCh38
rs968778063 Chr.3: 186852952-186852952 on Build GRCh38
rs199668131 Chr.3: 186853039-186853039 on Build GRCh38
rs201681868 Chr.3: 186853021-186853021 on Build GRCh38
rs940859604 Chr.3: 186852976-186852976 on Build GRCh38
rs780572021 Chr.3: 186853009-186853009 on Build GRCh38
rs562861639 Chr.3: 186852939-186852939 on Build GRCh38
rs2241766 Chr.3: 186853103-186853103 on Build GRCh38
rs905353312 Chr.3: 186852969-186852969 on Build GRCh38
rs766715892 Chr.3: 186853078-186853078 on Build GRCh38
rs537741070 Chr.3: 186853006-186853006 on Build GRCh38
rs956518635 Chr.3: 186852893-186852893 on Build GRCh38
rs747636727 Chr.3: 186853016-186853016 on Build GRCh38
rs181064565 Chr.3: 186852986-186852986 on Build GRCh38
rs17366653 Chr.3: 186853027-186853027 on Build GRCh38
rs192249161 Chr.3: 186852972-186852972 on Build GRCh38
rs765473337 Chr.3: 186853051-186853051 on Build GRCh38
rs994525210 Chr.3: 186853008-186853008 on Build GRCh38
rs36219758 Chr.3: 186852995-186852995 on Build GRCh38
rs200544481 Chr.3: 186853050-186853050 on Build GRCh38
rs202117803 Chr.3: 186853085-186853085 on Build GRCh38
rs376174575 Chr.3: 186853097-186853097 on Build GRCh38
rs777281402 Chr.3: 186853079-186853080 on Build GRCh38
rs199662617 Chr.3: 186853082-186853082 on Build GRCh38
rs776583777 Chr.3: 186853048-186853048 on Build GRCh38
rs774493377 Chr.3: 186853035-186853035 on Build GRCh38
rs922685531 Chr.3: 186852926-186852926 on Build GRCh38
rs547607723 Chr.3: 186853080-186853082 on Build GRCh38
rs749464930 Chr.3: 186853014-186853014 on Build GRCh38
rs201510126 Chr.3: 186853032-186853032 on Build GRCh38
rs551027621 Chr.3: 186852886-186852886 on Build GRCh38
rs1045241563 Chr.3: 186852968-186852968 on Build GRCh38
rs146334862 Chr.3: 186852966-186852966 on Build GRCh38
rs750611173 Chr.3: 186853052-186853052 on Build GRCh38
rs750132191 Chr.3: 186852971-186852971 on Build GRCh38
rs759747979 Chr.3: 186853080-186853081 on Build GRCh38
rs111455932 Chr.3: 186853049-186853049 on Build GRCh38
rs888701028 Chr.3: 186852940-186852940 on Build GRCh38
rs758856001 Chr.3: 186852928-186852928 on Build GRCh38
rs371911498 Chr.3: 186853038-186853038 on Build GRCh38
rs1018704392 Chr.3: 186853041-186853041 on Build GRCh38
rs201248773 Chr.3: 186853071-186853071 on Build GRCh38
rs537542748 Chr.3: 186853101-186853101 on Build GRCh38
rs757189922 Chr.3: 186853105-186853105 on Build GRCh38
rs17846866 Chr.3: 186852957-186852957 on Build GRCh38
rs777530847 Chr.3: 186853025-186853025 on Build GRCh38
rs567259133 Chr.3: 186853005-186853005 on Build GRCh38

View Full Product Details