Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58863683-58863928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Gene Symbol
C3orf67-AS1
Entrez Gene ID
101929238
Gene Name
C3orf67 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683953
Gene Chromosome Location
Chr.3: 58824471-59019093 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235776 c.1012C>T p.H338Y Chr.3: 58863764-58863764 on Build GRCh38
4941058 c.1075C>T p.P359S Chr.3: 58863701-58863701 on Build GRCh38
731782 c.1038G>A p.Q346Q Chr.3: 58863738-58863738 on Build GRCh38
6235777 c.1012C>T p.H338Y Chr.3: 58863764-58863764 on Build GRCh38
4870326 c.1063T>C p.F355L Chr.3: 58863713-58863713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986873649 Chr.3: 58863734-58863734 on Build GRCh38
rs760252763 Chr.3: 58863873-58863873 on Build GRCh38
rs201334935 Chr.3: 58863724-58863724 on Build GRCh38
rs768648328 Chr.3: 58863707-58863707 on Build GRCh38
rs750077560 Chr.3: 58863761-58863761 on Build GRCh38
rs182844803 Chr.3: 58863899-58863899 on Build GRCh38
rs751275835 Chr.3: 58863704-58863704 on Build GRCh38
rs749140287 Chr.3: 58863810-58863810 on Build GRCh38
rs779820032 Chr.3: 58863796-58863796 on Build GRCh38
rs773838863 Chr.3: 58863735-58863735 on Build GRCh38
rs112633530 Chr.3: 58863748-58863748 on Build GRCh38
rs772866007 Chr.3: 58863866-58863866 on Build GRCh38
rs568680728 Chr.3: 58863768-58863768 on Build GRCh38
rs747208104 Chr.3: 58863786-58863786 on Build GRCh38
rs536210537 Chr.3: 58863783-58863783 on Build GRCh38
rs758435990 Chr.3: 58863779-58863779 on Build GRCh38
rs765890334 Chr.3: 58863771-58863771 on Build GRCh38
rs35778488 Chr.3: 58863864-58863864 on Build GRCh38
rs777948716 Chr.3: 58863718-58863719 on Build GRCh38
rs747656672 Chr.3: 58863861-58863861 on Build GRCh38
rs752788408 Chr.3: 58863773-58863773 on Build GRCh38
rs774197666 Chr.3: 58863855-58863855 on Build GRCh38
rs780512640 Chr.3: 58863824-58863824 on Build GRCh38
rs112867020 Chr.3: 58863888-58863888 on Build GRCh38
rs755524466 Chr.3: 58863769-58863769 on Build GRCh38
rs757386555 Chr.3: 58863762-58863762 on Build GRCh38
rs768994756 Chr.3: 58863764-58863764 on Build GRCh38
rs927476169 Chr.3: 58863814-58863814 on Build GRCh38
rs766205138 Chr.3: 58863885-58863885 on Build GRCh38
rs147935364 Chr.3: 58863753-58863753 on Build GRCh38
rs761280367 Chr.3: 58863746-58863746 on Build GRCh38
rs141700320 Chr.3: 58863843-58863843 on Build GRCh38
rs755840434 Chr.3: 58863789-58863789 on Build GRCh38

View Full Product Details