Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24824317-24824590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
252327 c.2698G>A p.V900M Chr.2: 24824416-24824416 on Build GRCh38
1660274 c.2578-12_2578-9delCTGT p.? Chr.2: 24824545-24824548 on Build GRCh38
4539648 c.2715G>A p.L905L Chr.2: 24824399-24824399 on Build GRCh38
5526341 c.2635G>A p.E879K Chr.2: 24824479-24824479 on Build GRCh38
7094872 c.2603T>G p.F868C Chr.2: 24824511-24824511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201444015 Chr.2: 24824522-24824522 on Build GRCh38
rs983836313 Chr.2: 24824353-24824353 on Build GRCh38
rs7604576 Chr.2: 24824539-24824539 on Build GRCh38
rs376859524 Chr.2: 24824355-24824355 on Build GRCh38
rs762504497 Chr.2: 24824475-24824475 on Build GRCh38
rs769983667 Chr.2: 24824374-24824375 on Build GRCh38
rs372582116 Chr.2: 24824349-24824349 on Build GRCh38
rs770680230 Chr.2: 24824550-24824550 on Build GRCh38
rs748494944 Chr.2: 24824465-24824465 on Build GRCh38
rs199524620 Chr.2: 24824458-24824458 on Build GRCh38
rs967268038 Chr.2: 24824414-24824414 on Build GRCh38
rs780327182 Chr.2: 24824471-24824471 on Build GRCh38
rs772758883 Chr.2: 24824345-24824345 on Build GRCh38
rs750626685 Chr.2: 24824375-24824375 on Build GRCh38
rs771722608 Chr.2: 24824556-24824556 on Build GRCh38
rs770063939 Chr.2: 24824470-24824470 on Build GRCh38
rs751596841 Chr.2: 24824519-24824519 on Build GRCh38
rs201869480 Chr.2: 24824356-24824356 on Build GRCh38
rs374190273 Chr.2: 24824409-24824409 on Build GRCh38
rs963830964 Chr.2: 24824432-24824432 on Build GRCh38
rs763802630 Chr.2: 24824505-24824505 on Build GRCh38
rs1031082505 Chr.2: 24824450-24824450 on Build GRCh38
rs140258004 Chr.2: 24824417-24824417 on Build GRCh38
rs779149335 Chr.2: 24824374-24824374 on Build GRCh38
rs138203123 Chr.2: 24824498-24824498 on Build GRCh38
rs745325678 Chr.2: 24824555-24824555 on Build GRCh38
rs373554200 Chr.2: 24824551-24824551 on Build GRCh38
rs755894922 Chr.2: 24824366-24824366 on Build GRCh38
rs755070756 Chr.2: 24824527-24824527 on Build GRCh38
rs775433380 Chr.2: 24824565-24824565 on Build GRCh38
rs762464507 Chr.2: 24824347-24824347 on Build GRCh38
rs752601873 Chr.2: 24824491-24824491 on Build GRCh38
rs370437393 Chr.2: 24824546-24824546 on Build GRCh38
rs369517755 Chr.2: 24824476-24824476 on Build GRCh38
rs773980429 Chr.2: 24824488-24824488 on Build GRCh38
rs765988437 Chr.2: 24824348-24824348 on Build GRCh38
rs529625336 Chr.2: 24824542-24824542 on Build GRCh38
rs540964272 Chr.2: 24824489-24824489 on Build GRCh38
rs374101291 Chr.2: 24824507-24824507 on Build GRCh38
rs952191315 Chr.2: 24824350-24824350 on Build GRCh38
rs989488388 Chr.2: 24824487-24824487 on Build GRCh38
rs373810353 Chr.2: 24824562-24824562 on Build GRCh38
rs758436691 Chr.2: 24824405-24824405 on Build GRCh38
rs760462078 Chr.2: 24824499-24824499 on Build GRCh38
rs756707162 Chr.2: 24824544-24824544 on Build GRCh38
rs769445908 Chr.2: 24824343-24824343 on Build GRCh38
rs758597196 Chr.2: 24824508-24824508 on Build GRCh38
rs766627903 Chr.2: 24824516-24824516 on Build GRCh38
rs529229449 Chr.2: 24824530-24824530 on Build GRCh38
rs769120427 Chr.2: 24824449-24824449 on Build GRCh38

View Full Product Details