Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22181438-22181671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
226358 c.1358_1359CC>TT p.T453>? Chr.18: 22181508-22181509 on Build GRCh38
986960 c.1367G>A p.R456H Chr.18: 22181517-22181517 on Build GRCh38
6003268 c.1309T>C p.S437P Chr.18: 22181459-22181459 on Build GRCh38
5633079 c.1378G>A p.E460K Chr.18: 22181528-22181528 on Build GRCh38
6030828 c.1407A>G p.G469G Chr.18: 22181557-22181557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768353106 Chr.18: 22181610-22181610 on Build GRCh38
rs934497510 Chr.18: 22181488-22181488 on Build GRCh38
rs751665524 Chr.18: 22181587-22181587 on Build GRCh38
rs765639422 Chr.18: 22181527-22181527 on Build GRCh38
rs762777966 Chr.18: 22181632-22181632 on Build GRCh38
rs989904962 Chr.18: 22181491-22181491 on Build GRCh38
rs765926114 Chr.18: 22181523-22181523 on Build GRCh38
rs781296792 Chr.18: 22181605-22181605 on Build GRCh38
rs387906817 Chr.18: 22181504-22181504 on Build GRCh38
rs151176879 Chr.18: 22181525-22181525 on Build GRCh38
rs761683725 Chr.18: 22181499-22181499 on Build GRCh38
rs387906818 Chr.18: 22181516-22181516 on Build GRCh38
rs770436209 Chr.18: 22181629-22181629 on Build GRCh38
rs772705320 Chr.18: 22181512-22181512 on Build GRCh38
rs61731977 Chr.18: 22181500-22181500 on Build GRCh38
rs768460903 Chr.18: 22181479-22181479 on Build GRCh38
rs774280237 Chr.18: 22181492-22181492 on Build GRCh38
rs143026087 Chr.18: 22181524-22181524 on Build GRCh38
rs746039582 Chr.18: 22181607-22181607 on Build GRCh38
rs387906819 Chr.18: 22181517-22181517 on Build GRCh38
rs777984822 Chr.18: 22181463-22181463 on Build GRCh38
rs757309596 Chr.18: 22181600-22181600 on Build GRCh38
rs897375080 Chr.18: 22181637-22181637 on Build GRCh38
rs387906820 Chr.18: 22181549-22181549 on Build GRCh38
rs771024527 Chr.18: 22181467-22181467 on Build GRCh38
rs387906813 Chr.18: 22181546-22181546 on Build GRCh38
rs146624465 Chr.18: 22181470-22181470 on Build GRCh38
rs199592618 Chr.18: 22181617-22181617 on Build GRCh38
rs758967085 Chr.18: 22181539-22181539 on Build GRCh38
rs368310322 Chr.18: 22181611-22181611 on Build GRCh38
rs747300699 Chr.18: 22181464-22181464 on Build GRCh38
rs778036747 Chr.18: 22181548-22181548 on Build GRCh38
rs772760319 Chr.18: 22181620-22181620 on Build GRCh38
rs200556553 Chr.18: 22181618-22181618 on Build GRCh38
rs760125896 Chr.18: 22181627-22181627 on Build GRCh38

View Full Product Details