Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37718962-37719235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
UniGene
Hs.163990
Gene Chromosome Location
Chr.9: 37602979-37746904 on Build GRCh38
Transcripts
NM_014907.2 XM_011517803.2 XM_011517804.2 XM_011517805.2 XM_011517806.2 XM_017014480.1 XM_017014481.1 XM_017014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1701095 c.505_506GG>AA p.G169K Chr.9: 37719165-37719166 on Build GRCh38
1109019 c.441G>T p.E147D Chr.9: 37719101-37719101 on Build GRCh38
3657409 c.45G>A p.E15E Chr.9: 37719098-37719098 on Build GRCh38
1238942 c.435C>T p.T145T Chr.9: 37719095-37719095 on Build GRCh38
3657408 c.438G>A p.E146E Chr.9: 37719098-37719098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751418524 Chr.9: 37719084-37719084 on Build GRCh38
rs765067 Chr.9: 37719104-37719104 on Build GRCh38
rs754712459 Chr.9: 37719094-37719094 on Build GRCh38
rs376402611 Chr.9: 37719071-37719071 on Build GRCh38
rs373549069 Chr.9: 37719046-37719046 on Build GRCh38
rs769184268 Chr.9: 37719181-37719181 on Build GRCh38
rs756042056 Chr.9: 37719129-37719129 on Build GRCh38
rs752583725 Chr.9: 37719125-37719125 on Build GRCh38
rs2013458 Chr.9: 37719014-37719014 on Build GRCh38
rs182195690 Chr.9: 37718998-37718998 on Build GRCh38
rs202228676 Chr.9: 37719175-37719175 on Build GRCh38
rs775848929 Chr.9: 37719057-37719057 on Build GRCh38
rs866518755 Chr.9: 37719108-37719108 on Build GRCh38
rs775342241 Chr.9: 37719000-37719017 on Build GRCh38
rs369706572 Chr.9: 37719096-37719096 on Build GRCh38
rs559954392 Chr.9: 37719082-37719082 on Build GRCh38
rs774428491 Chr.9: 37719095-37719095 on Build GRCh38
rs369468677 Chr.9: 37719135-37719135 on Build GRCh38
rs772704712 Chr.9: 37719053-37719053 on Build GRCh38
rs543341119 Chr.9: 37719017-37719017 on Build GRCh38
rs554335232 Chr.9: 37719001-37719001 on Build GRCh38
rs375958396 Chr.9: 37719215-37719215 on Build GRCh38
rs746303862 Chr.9: 37719167-37719167 on Build GRCh38
rs866778035 Chr.9: 37719165-37719165 on Build GRCh38
rs777348648 Chr.9: 37719096-37719098 on Build GRCh38
rs765980830 Chr.9: 37719083-37719083 on Build GRCh38
rs772745763 Chr.9: 37719193-37719193 on Build GRCh38
rs779275445 Chr.9: 37719162-37719162 on Build GRCh38
rs764683777 Chr.9: 37719080-37719080 on Build GRCh38
rs751222485 Chr.9: 37719123-37719123 on Build GRCh38
rs757594781 Chr.9: 37719154-37719154 on Build GRCh38
rs775883026 Chr.9: 37719173-37719173 on Build GRCh38
rs1020684720 Chr.9: 37719019-37719019 on Build GRCh38
rs868560965 Chr.9: 37719186-37719186 on Build GRCh38
rs761175613 Chr.9: 37719070-37719070 on Build GRCh38
rs372943040 Chr.9: 37719210-37719210 on Build GRCh38
rs777935741 Chr.9: 37719032-37719032 on Build GRCh38
rs377512929 Chr.9: 37719216-37719216 on Build GRCh38
rs772326418 Chr.9: 37719168-37719168 on Build GRCh38
rs759949376 Chr.9: 37719049-37719049 on Build GRCh38
rs778026562 Chr.9: 37719138-37719138 on Build GRCh38
rs771233733 Chr.9: 37719036-37719036 on Build GRCh38
rs966336839 Chr.9: 37719037-37719037 on Build GRCh38
rs749357864 Chr.9: 37719139-37719139 on Build GRCh38
rs771117077 Chr.9: 37719091-37719091 on Build GRCh38
rs749556531 Chr.9: 37719035-37719035 on Build GRCh38

View Full Product Details