Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17735955-17736204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
361556 c.151C>A p.R51S Chr.11: 17736153-17736153 on Build GRCh38
5492768 c.53G>A p.R18H Chr.11: 17736055-17736055 on Build GRCh38
4145604 c.99G>T p.R33R Chr.11: 17736101-17736101 on Build GRCh38
4091269 c.123C>T p.D41D Chr.11: 17736125-17736125 on Build GRCh38
6690579 c.152G>A p.R51H Chr.11: 17736154-17736154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762388292 Chr.11: 17736126-17736126 on Build GRCh38
rs200559511 Chr.11: 17736130-17736130 on Build GRCh38
rs200677353 Chr.11: 17736116-17736116 on Build GRCh38
rs929174953 Chr.11: 17735971-17735971 on Build GRCh38
rs140637569 Chr.11: 17736098-17736098 on Build GRCh38
rs370742331 Chr.11: 17736104-17736104 on Build GRCh38
rs766667950 Chr.11: 17735992-17735992 on Build GRCh38
rs766715132 Chr.11: 17736154-17736154 on Build GRCh38
rs762218553 Chr.11: 17736080-17736080 on Build GRCh38
rs763076681 Chr.11: 17736150-17736150 on Build GRCh38
rs778368338 Chr.11: 17736101-17736101 on Build GRCh38
rs993728478 Chr.11: 17736062-17736062 on Build GRCh38
rs777058455 Chr.11: 17736118-17736118 on Build GRCh38
rs777640256 Chr.11: 17736061-17736061 on Build GRCh38
rs747336979 Chr.11: 17736115-17736115 on Build GRCh38
rs755872953 Chr.11: 17736028-17736028 on Build GRCh38
rs529678348 Chr.11: 17735972-17735972 on Build GRCh38
rs773455390 Chr.11: 17736133-17736133 on Build GRCh38
rs752454923 Chr.11: 17736002-17736002 on Build GRCh38
rs181020482 Chr.11: 17736035-17736035 on Build GRCh38
rs763235319 Chr.11: 17735978-17735978 on Build GRCh38
rs76929858 Chr.11: 17736074-17736074 on Build GRCh38
rs779700926 Chr.11: 17736105-17736105 on Build GRCh38

View Full Product Details