Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80888600-80888873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEMIP
Entrez Gene ID
57214
Gene Name
cell migration inducing hyaluronan binding protein
Gene Aliases
CCSP1, HYBID, KIAA1199, TMEM2L
UniGene
Hs.459088
Gene Chromosome Location
Chr.15: 80779343-80951662 on Build GRCh38
Transcripts
NM_001293298.1 NM_001293304.1 NM_018689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077956 c.916G>C p.E306Q Chr.15: 80888748-80888748 on Build GRCh38
4770300 c.920A>G p.H307R Chr.15: 80888752-80888752 on Build GRCh38
7317946 c.900C>G p.F300L Chr.15: 80888732-80888732 on Build GRCh38
965525 c.921T>C p.H307H Chr.15: 80888753-80888753 on Build GRCh38
4774326 c.934A>G p.N312D Chr.15: 80888766-80888766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8028511 Chr.15: 80888640-80888640 on Build GRCh38
rs764306463 Chr.15: 80888773-80888773 on Build GRCh38
rs1007177974 Chr.15: 80888756-80888756 on Build GRCh38
rs759087523 Chr.15: 80888757-80888757 on Build GRCh38
rs377578871 Chr.15: 80888767-80888767 on Build GRCh38
rs891487354 Chr.15: 80888651-80888651 on Build GRCh38
rs74030632 Chr.15: 80888624-80888624 on Build GRCh38
rs373591542 Chr.15: 80888753-80888753 on Build GRCh38
rs749543042 Chr.15: 80888696-80888696 on Build GRCh38
rs771721238 Chr.15: 80888706-80888706 on Build GRCh38
rs151251075 Chr.15: 80888783-80888783 on Build GRCh38
rs550082260 Chr.15: 80888724-80888724 on Build GRCh38
rs370075183 Chr.15: 80888812-80888812 on Build GRCh38
rs757774341 Chr.15: 80888669-80888669 on Build GRCh38
rs780414605 Chr.15: 80888687-80888687 on Build GRCh38
rs950747379 Chr.15: 80888831-80888831 on Build GRCh38
rs1051319586 Chr.15: 80888709-80888709 on Build GRCh38
rs781706148 Chr.15: 80888672-80888672 on Build GRCh38
rs751938826 Chr.15: 80888777-80888777 on Build GRCh38
rs868160183 Chr.15: 80888700-80888700 on Build GRCh38
rs201258656 Chr.15: 80888797-80888797 on Build GRCh38
rs980292011 Chr.15: 80888620-80888620 on Build GRCh38
rs780210530 Chr.15: 80888821-80888821 on Build GRCh38
rs769899499 Chr.15: 80888685-80888685 on Build GRCh38
rs754830713 Chr.15: 80888769-80888769 on Build GRCh38
rs749400482 Chr.15: 80888829-80888829 on Build GRCh38
rs201049604 Chr.15: 80888725-80888725 on Build GRCh38
rs746420704 Chr.15: 80888802-80888802 on Build GRCh38
rs773067476 Chr.15: 80888720-80888720 on Build GRCh38
rs896191037 Chr.15: 80888636-80888636 on Build GRCh38
rs936246771 Chr.15: 80888632-80888632 on Build GRCh38
rs146995917 Chr.15: 80888743-80888743 on Build GRCh38

View Full Product Details