Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50751837-50752074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLD
Entrez Gene ID
1540
Gene Name
CYLD lysine 63 deubiquitinase
Gene Aliases
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2
UniGene
Hs.578973
Gene Chromosome Location
Chr.16: 50742026-50801935 on Build GRCh38
Transcripts
NM_001042355.1 NM_001042412.1 NM_015247.2 XM_005255812.2 XM_006721149.1 XM_011522907.2 XM_011522908.1 XM_011522911.2 XM_017022977.1 XM_017022978.1 XM_017022979.1 XM_017022980.1 XM_017022981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7081287 c.757G>A p.D253N Chr.16: 50751856-50751856 on Build GRCh38
7081286 c.757G>A p.D253N Chr.16: 50751856-50751856 on Build GRCh38
4061127 c.775G>A p.E259K Chr.16: 50751874-50751874 on Build GRCh38
5614366 c.802G>T p.D268Y Chr.16: 50751901-50751901 on Build GRCh38
5614365 c.802G>T p.D268Y Chr.16: 50751901-50751901 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1036323857 Chr.16: 50751966-50751966 on Build GRCh38
rs899777762 Chr.16: 50751968-50751968 on Build GRCh38
rs537111724 Chr.16: 50751892-50751892 on Build GRCh38
rs899911468 Chr.16: 50752010-50752028 on Build GRCh38
rs867267998 Chr.16: 50751865-50751865 on Build GRCh38
rs66613485 Chr.16: 50751980-50751993 on Build GRCh38
rs994953668 Chr.16: 50751997-50751997 on Build GRCh38
rs373370558 Chr.16: 50751979-50751979 on Build GRCh38
rs370534101 Chr.16: 50752003-50752003 on Build GRCh38
rs778588431 Chr.16: 50751964-50751965 on Build GRCh38
rs1039677518 Chr.16: 50751965-50751966 on Build GRCh38
rs769304456 Chr.16: 50751928-50751928 on Build GRCh38
rs376859556 Chr.16: 50751942-50751942 on Build GRCh38
rs897918884 Chr.16: 50751976-50751976 on Build GRCh38
rs903569210 Chr.16: 50752014-50752014 on Build GRCh38
rs777590103 Chr.16: 50751946-50751949 on Build GRCh38
rs747446180 Chr.16: 50751922-50751922 on Build GRCh38
rs748884735 Chr.16: 50751952-50751953 on Build GRCh38
rs761539625 Chr.16: 50751939-50751939 on Build GRCh38
rs775035809 Chr.16: 50751930-50751930 on Build GRCh38
rs57372743 Chr.16: 50751993-50751993 on Build GRCh38
rs941364161 Chr.16: 50751959-50751959 on Build GRCh38
rs57988572 Chr.16: 50751991-50751991 on Build GRCh38
rs892336849 Chr.16: 50751984-50751995 on Build GRCh38
rs540689897 Chr.16: 50751934-50751934 on Build GRCh38
rs763696277 Chr.16: 50751932-50751932 on Build GRCh38
rs532396672 Chr.16: 50752037-50752037 on Build GRCh38
rs868007802 Chr.16: 50751996-50751996 on Build GRCh38
rs773736598 Chr.16: 50751921-50751921 on Build GRCh38
rs772431475 Chr.16: 50751900-50751900 on Build GRCh38
rs762523768 Chr.16: 50751931-50751931 on Build GRCh38
rs748510262 Chr.16: 50751876-50751876 on Build GRCh38
rs11271535 Chr.16: 50752004-50752017 on Build GRCh38
rs1027734059 Chr.16: 50752012-50752012 on Build GRCh38
rs750391169 Chr.16: 50751952-50751952 on Build GRCh38
rs770259102 Chr.16: 50751937-50751938 on Build GRCh38

View Full Product Details