Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35119322-35119547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
UniGene
Hs.301350
Gene Chromosome Location
Chr.19: 35115821-35124324 on Build GRCh38
Transcripts
NM_001136007.1 NM_001136008.1 NM_001136009.1 NM_001136010.1 NM_001136011.1 NM_001136012.1 NM_005971.3 NM_021910.2 XM_005258996.4 XM_011527032.2 XM_017026877.1 XM_017026878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7169593 c.169G>A p.D57N Chr.19: 35119374-35119374 on Build GRCh38
6049530 c.179A>G p.K60R Chr.19: 35119384-35119384 on Build GRCh38
1161269 c.9G>T p.K3N Chr.19: 35119385-35119385 on Build GRCh38
6667991 c.15C>T p.T5T Chr.19: 35119391-35119391 on Build GRCh38
6667992 c.15C>T p.T5T Chr.19: 35119391-35119391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779383283 Chr.19: 35119403-35119403 on Build GRCh38
rs747522387 Chr.19: 35119464-35119465 on Build GRCh38
rs780778533 Chr.19: 35119354-35119354 on Build GRCh38
rs745372389 Chr.19: 35119357-35119357 on Build GRCh38
rs933047292 Chr.19: 35119340-35119340 on Build GRCh38
rs2278994 Chr.19: 35119341-35119341 on Build GRCh38
rs746593023 Chr.19: 35119369-35119369 on Build GRCh38
rs769131598 Chr.19: 35119359-35119359 on Build GRCh38
rs779622390 Chr.19: 35119363-35119363 on Build GRCh38
rs903043794 Chr.19: 35119507-35119507 on Build GRCh38
rs763423896 Chr.19: 35119380-35119380 on Build GRCh38
rs145627296 Chr.19: 35119398-35119398 on Build GRCh38
rs749200090 Chr.19: 35119364-35119364 on Build GRCh38
rs751966616 Chr.19: 35119350-35119350 on Build GRCh38
rs370666338 Chr.19: 35119433-35119434 on Build GRCh38
rs770100820 Chr.19: 35119464-35119464 on Build GRCh38
rs746026649 Chr.19: 35119461-35119461 on Build GRCh38
rs919425174 Chr.19: 35119339-35119339 on Build GRCh38
rs754961156 Chr.19: 35119421-35119421 on Build GRCh38
rs370586631 Chr.19: 35119349-35119349 on Build GRCh38
rs540951596 Chr.19: 35119508-35119508 on Build GRCh38
rs748858058 Chr.19: 35119405-35119405 on Build GRCh38
rs752841610 Chr.19: 35119377-35119377 on Build GRCh38
rs933912031 Chr.19: 35119478-35119478 on Build GRCh38
rs756875089 Chr.19: 35119352-35119352 on Build GRCh38
rs751921647 Chr.19: 35119388-35119388 on Build GRCh38
rs757679976 Chr.19: 35119391-35119391 on Build GRCh38
rs768737517 Chr.19: 35119368-35119368 on Build GRCh38
rs112543884 Chr.19: 35119418-35119418 on Build GRCh38
rs765187472 Chr.19: 35119376-35119376 on Build GRCh38
rs1041606937 Chr.19: 35119505-35119505 on Build GRCh38
rs376810232 Chr.19: 35119434-35119434 on Build GRCh38
rs371019682 Chr.19: 35119343-35119343 on Build GRCh38
rs773087908 Chr.19: 35119458-35119458 on Build GRCh38
rs755824659 Chr.19: 35119399-35119399 on Build GRCh38
rs62120434 Chr.19: 35119384-35119384 on Build GRCh38
rs781632442 Chr.19: 35119396-35119396 on Build GRCh38
rs964926998 Chr.19: 35119524-35119524 on Build GRCh38
rs560518984 Chr.19: 35119523-35119523 on Build GRCh38
rs1032630967 Chr.19: 35119491-35119491 on Build GRCh38
rs775512792 Chr.19: 35119465-35119465 on Build GRCh38
rs768386918 Chr.19: 35119370-35119370 on Build GRCh38
rs779050924 Chr.19: 35119433-35119433 on Build GRCh38
rs575016663 Chr.19: 35119498-35119498 on Build GRCh38
rs979426193 Chr.19: 35119488-35119488 on Build GRCh38
rs771696799 Chr.19: 35119445-35119445 on Build GRCh38
rs369663190 Chr.19: 35119475-35119475 on Build GRCh38
rs776657302 Chr.19: 35119372-35119372 on Build GRCh38
rs113461255 Chr.19: 35119485-35119485 on Build GRCh38

View Full Product Details