Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161692080-161692305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA6
Entrez Gene ID
2559
Gene Name
gamma-aminobutyric acid type A receptor alpha6 subunit
Gene Aliases
-
UniGene
Hs.90791
Gene Chromosome Location
Chr.5: 161685652-161702592 on Build GRCh38
Transcripts
NM_000811.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4422433 c.1086+8T>C p.? Chr.5: 161692208-161692208 on Build GRCh38
4989088 c.1053A>C p.K351N Chr.5: 161692167-161692167 on Build GRCh38
268058 c.1033C>T p.P345S Chr.5: 161692147-161692147 on Build GRCh38
7167859 c.1035C>G p.P345P Chr.5: 161692149-161692149 on Build GRCh38
3827613 c.1075G>A p.E359K Chr.5: 161692189-161692189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141165453 Chr.5: 161692210-161692210 on Build GRCh38
rs765780675 Chr.5: 161692120-161692120 on Build GRCh38
rs759363086 Chr.5: 161692145-161692145 on Build GRCh38
rs755988030 Chr.5: 161692161-161692161 on Build GRCh38
rs773896796 Chr.5: 161692202-161692202 on Build GRCh38
rs376973590 Chr.5: 161692178-161692178 on Build GRCh38
rs79970352 Chr.5: 161692270-161692270 on Build GRCh38
rs759042199 Chr.5: 161692231-161692231 on Build GRCh38
rs540538801 Chr.5: 161692243-161692243 on Build GRCh38
rs767281233 Chr.5: 161692149-161692149 on Build GRCh38
rs149661341 Chr.5: 161692238-161692238 on Build GRCh38
rs142221692 Chr.5: 161692181-161692181 on Build GRCh38
rs539296903 Chr.5: 161692173-161692173 on Build GRCh38
rs766433512 Chr.5: 161692245-161692245 on Build GRCh38
rs750458393 Chr.5: 161692167-161692167 on Build GRCh38
rs766023505 Chr.5: 161692133-161692133 on Build GRCh38
rs748784879 Chr.5: 161692208-161692208 on Build GRCh38
rs13184586 Chr.5: 161692119-161692119 on Build GRCh38
rs773660339 Chr.5: 161692193-161692193 on Build GRCh38
rs1001887112 Chr.5: 161692146-161692146 on Build GRCh38
rs751085820 Chr.5: 161692238-161692238 on Build GRCh38
rs760494092 Chr.5: 161692236-161692236 on Build GRCh38
rs144044129 Chr.5: 161692125-161692125 on Build GRCh38
rs774050094 Chr.5: 161692211-161692211 on Build GRCh38
rs956200041 Chr.5: 161692269-161692269 on Build GRCh38
rs773925965 Chr.5: 161692138-161692138 on Build GRCh38
rs755235727 Chr.5: 161692177-161692177 on Build GRCh38
rs747249350 Chr.5: 161692176-161692176 on Build GRCh38
rs920562311 Chr.5: 161692275-161692275 on Build GRCh38
rs762584200 Chr.5: 161692118-161692118 on Build GRCh38
rs758508674 Chr.5: 161692172-161692172 on Build GRCh38
rs377668354 Chr.5: 161692233-161692233 on Build GRCh38
rs770387928 Chr.5: 161692194-161692194 on Build GRCh38
rs1011807163 Chr.5: 161692271-161692271 on Build GRCh38
rs76937628 Chr.5: 161692154-161692154 on Build GRCh38
rs202216005 Chr.5: 161692148-161692148 on Build GRCh38
rs751675134 Chr.5: 161692247-161692247 on Build GRCh38
rs763985245 Chr.5: 161692237-161692237 on Build GRCh38
rs764040523 Chr.5: 161692162-161692162 on Build GRCh38

View Full Product Details