Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143726825-143727080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83H
Entrez Gene ID
286077
Gene Name
family with sequence similarity 83 member H
Gene Aliases
AI3
UniGene
Hs.67776
Gene Chromosome Location
Chr.8: 143723933-143739947 on Build GRCh38
Transcripts
NM_198488.3 XM_005250887.3 XM_005250888.3 XM_005250889.3 XM_011516980.2 XM_011516981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4762285 c.2547C>G p.D849E Chr.8: 143726914-143726914 on Build GRCh38
7299049 c.2447C>T p.A816V Chr.8: 143727014-143727014 on Build GRCh38
6355782 c.2610C>T p.S870S Chr.8: 143726851-143726851 on Build GRCh38
4433933 c.2438C>T p.S813L Chr.8: 143727023-143727023 on Build GRCh38
3395149 c.2410G>T p.E804* Chr.8: 143727051-143727051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782023240 Chr.8: 143726972-143726972 on Build GRCh38
rs782747990 Chr.8: 143726971-143726971 on Build GRCh38
rs782409778 Chr.8: 143726889-143726889 on Build GRCh38
rs782330598 Chr.8: 143727035-143727035 on Build GRCh38
rs782626750 Chr.8: 143726934-143726934 on Build GRCh38
rs781947847 Chr.8: 143726967-143726967 on Build GRCh38
rs782554162 Chr.8: 143726850-143726850 on Build GRCh38
rs781886472 Chr.8: 143726848-143726848 on Build GRCh38
rs373496375 Chr.8: 143726903-143726903 on Build GRCh38
rs372293894 Chr.8: 143726902-143726902 on Build GRCh38
rs781983772 Chr.8: 143727028-143727028 on Build GRCh38
rs782616861 Chr.8: 143727013-143727013 on Build GRCh38
rs782359952 Chr.8: 143726898-143726898 on Build GRCh38
rs529733873 Chr.8: 143726909-143726909 on Build GRCh38
rs782066813 Chr.8: 143726973-143726973 on Build GRCh38
rs782471637 Chr.8: 143726987-143726987 on Build GRCh38
rs781849797 Chr.8: 143726983-143726983 on Build GRCh38
rs782444873 Chr.8: 143726930-143726930 on Build GRCh38
rs530393090 Chr.8: 143727000-143727000 on Build GRCh38
rs1054886527 Chr.8: 143726871-143726871 on Build GRCh38
rs781925162 Chr.8: 143727041-143727041 on Build GRCh38
rs782285804 Chr.8: 143727034-143727034 on Build GRCh38
rs1017685992 Chr.8: 143726947-143726947 on Build GRCh38
rs781831914 Chr.8: 143726854-143726854 on Build GRCh38
rs782647191 Chr.8: 143726945-143726945 on Build GRCh38
rs782430933 Chr.8: 143727011-143727011 on Build GRCh38
rs782294305 Chr.8: 143726951-143726951 on Build GRCh38
rs782727032 Chr.8: 143727009-143727010 on Build GRCh38
rs782192124 Chr.8: 143727007-143727007 on Build GRCh38
rs1028257176 Chr.8: 143726996-143726996 on Build GRCh38
rs990521598 Chr.8: 143727052-143727052 on Build GRCh38
rs782353953 Chr.8: 143726961-143726961 on Build GRCh38
rs782653370 Chr.8: 143726870-143726870 on Build GRCh38
rs782215806 Chr.8: 143726941-143726941 on Build GRCh38
rs782209001 Chr.8: 143727016-143727016 on Build GRCh38
rs367570466 Chr.8: 143726883-143726883 on Build GRCh38
rs782736726 Chr.8: 143726926-143726926 on Build GRCh38
rs200587785 Chr.8: 143726956-143726956 on Build GRCh38
rs532569154 Chr.8: 143726868-143726868 on Build GRCh38
rs552468993 Chr.8: 143727059-143727059 on Build GRCh38
rs782089523 Chr.8: 143726918-143726918 on Build GRCh38
rs781933799 Chr.8: 143726955-143726955 on Build GRCh38
rs374727482 Chr.8: 143726845-143726845 on Build GRCh38
rs782708415 Chr.8: 143726974-143726974 on Build GRCh38
rs782654478 Chr.8: 143727004-143727004 on Build GRCh38
rs376843775 Chr.8: 143726927-143726927 on Build GRCh38
rs1008896900 Chr.8: 143726938-143726938 on Build GRCh38
rs782514192 Chr.8: 143726943-143726943 on Build GRCh38
rs369310812 Chr.8: 143726894-143726894 on Build GRCh38
rs902408587 Chr.8: 143726922-143726922 on Build GRCh38
rs781984260 Chr.8: 143726893-143726893 on Build GRCh38
rs782125165 Chr.8: 143726968-143726968 on Build GRCh38
rs916321861 Chr.8: 143727062-143727062 on Build GRCh38
rs782455374 Chr.8: 143726849-143726849 on Build GRCh38
rs782104094 Chr.8: 143727047-143727047 on Build GRCh38
rs781812004 Chr.8: 143726929-143726929 on Build GRCh38
rs781790565 Chr.8: 143726910-143726910 on Build GRCh38
rs375864628 Chr.8: 143726890-143726890 on Build GRCh38
rs782033920 Chr.8: 143726907-143726907 on Build GRCh38
rs370579237 Chr.8: 143726884-143726884 on Build GRCh38
rs890462111 Chr.8: 143726932-143726932 on Build GRCh38
rs782415634 Chr.8: 143726953-143726953 on Build GRCh38

View Full Product Details