Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 130163361-130163600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD10
Entrez Gene ID
11211
Gene Name
frizzled class receptor 10
Gene Aliases
CD350, FZ-10, Fz10, FzE7, hFz10
UniGene
Hs.31664
Gene Chromosome Location
Chr.12: 130162459-130165740 on Build GRCh38
Transcripts
NM_007197.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3458069 c.549G>A p.K183K Chr.12: 130163491-130163491 on Build GRCh38
5034079 c.524A>C p.H175P Chr.12: 130163466-130163466 on Build GRCh38
936963 c.648C>T p.D216D Chr.12: 130163590-130163590 on Build GRCh38
1207709 c.613G>A p.A205T Chr.12: 130163555-130163555 on Build GRCh38
4639845 c.569G>A p.G190D Chr.12: 130163511-130163511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370632753 Chr.12: 130163553-130163553 on Build GRCh38
rs756092983 Chr.12: 130163427-130163427 on Build GRCh38
rs756870051 Chr.12: 130163413-130163413 on Build GRCh38
rs765524304 Chr.12: 130163484-130163484 on Build GRCh38
rs563264151 Chr.12: 130163464-130163464 on Build GRCh38
rs985841216 Chr.12: 130163402-130163402 on Build GRCh38
rs892018995 Chr.12: 130163450-130163450 on Build GRCh38
rs765136161 Chr.12: 130163564-130163564 on Build GRCh38
rs777914537 Chr.12: 130163503-130163503 on Build GRCh38
rs752522464 Chr.12: 130163569-130163569 on Build GRCh38
rs964564427 Chr.12: 130163410-130163410 on Build GRCh38
rs143887209 Chr.12: 130163404-130163404 on Build GRCh38
rs759948678 Chr.12: 130163406-130163406 on Build GRCh38
rs973296981 Chr.12: 130163428-130163428 on Build GRCh38
rs777626340 Chr.12: 130163581-130163581 on Build GRCh38
rs767188838 Chr.12: 130163533-130163533 on Build GRCh38
rs763279090 Chr.12: 130163528-130163528 on Build GRCh38
rs775744371 Chr.12: 130163493-130163493 on Build GRCh38
rs201661032 Chr.12: 130163498-130163498 on Build GRCh38
rs774773609 Chr.12: 130163390-130163390 on Build GRCh38
rs1030287197 Chr.12: 130163440-130163440 on Build GRCh38
rs761795300 Chr.12: 130163391-130163391 on Build GRCh38
rs749936930 Chr.12: 130163424-130163424 on Build GRCh38
rs767118059 Chr.12: 130163495-130163495 on Build GRCh38
rs773187403 Chr.12: 130163395-130163395 on Build GRCh38
rs1017978465 Chr.12: 130163530-130163530 on Build GRCh38
rs768979118 Chr.12: 130163383-130163383 on Build GRCh38
rs772214243 Chr.12: 130163443-130163443 on Build GRCh38
rs761203938 Chr.12: 130163398-130163398 on Build GRCh38
rs770928042 Chr.12: 130163507-130163507 on Build GRCh38
rs773617704 Chr.12: 130163495-130163495 on Build GRCh38
rs778729120 Chr.12: 130163499-130163499 on Build GRCh38
rs753422384 Chr.12: 130163556-130163556 on Build GRCh38
rs747106141 Chr.12: 130163449-130163449 on Build GRCh38
rs939134736 Chr.12: 130163506-130163506 on Build GRCh38
rs772807968 Chr.12: 130163543-130163543 on Build GRCh38
rs749084019 Chr.12: 130163436-130163436 on Build GRCh38
rs771412434 Chr.12: 130163455-130163455 on Build GRCh38
rs772122832 Chr.12: 130163392-130163392 on Build GRCh38
rs776701558 Chr.12: 130163511-130163511 on Build GRCh38
rs965418997 Chr.12: 130163532-130163532 on Build GRCh38
rs755745804 Chr.12: 130163497-130163497 on Build GRCh38
rs754888654 Chr.12: 130163437-130163437 on Build GRCh38
rs759513875 Chr.12: 130163559-130163559 on Build GRCh38
rs763728060 Chr.12: 130163411-130163411 on Build GRCh38
rs747933777 Chr.12: 130163501-130163501 on Build GRCh38
rs758622904 Chr.12: 130163502-130163502 on Build GRCh38
rs770320717 Chr.12: 130163517-130163517 on Build GRCh38
rs766910008 Chr.12: 130163401-130163401 on Build GRCh38
rs777186624 Chr.12: 130163460-130163460 on Build GRCh38
rs780192582 Chr.12: 130163432-130163432 on Build GRCh38
rs374447637 Chr.12: 130163451-130163451 on Build GRCh38
rs746320895 Chr.12: 130163512-130163512 on Build GRCh38
rs767008901 Chr.12: 130163417-130163417 on Build GRCh38
rs747084549 Chr.12: 130163505-130163505 on Build GRCh38
rs775897205 Chr.12: 130163526-130163526 on Build GRCh38
rs748345546 Chr.12: 130163442-130163442 on Build GRCh38
rs758192337 Chr.12: 130163578-130163578 on Build GRCh38
rs765929480 Chr.12: 130163544-130163544 on Build GRCh38
rs549217660 Chr.12: 130163582-130163582 on Build GRCh38
rs543498086 Chr.12: 130163439-130163439 on Build GRCh38

View Full Product Details