Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 192143916-192144189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF12
Entrez Gene ID
2257
Gene Name
fibroblast growth factor 12
Gene Aliases
FGF12B, FHF1
UniGene
Hs.390250
Gene Chromosome Location
Chr.3: 192139393-192727599 on Build GRCh38
Transcripts
NM_004113.5 NM_021032.4 XM_005247227.2 XM_006713538.3 XM_017005879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3735254 c.302C>T p.T101I Chr.3: 192144055-192144055 on Build GRCh38
6663477 c.479G>A p.R160H Chr.3: 192144076-192144076 on Build GRCh38
4116078 c.478C>T p.R160C Chr.3: 192144077-192144077 on Build GRCh38
7147601 c.296C>G p.S99C Chr.3: 192144061-192144061 on Build GRCh38
1042560 c.252G>A p.S84S Chr.3: 192144105-192144105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962617533 Chr.3: 192144072-192144072 on Build GRCh38
rs762062680 Chr.3: 192144149-192144149 on Build GRCh38
rs747782511 Chr.3: 192144083-192144083 on Build GRCh38
rs552996417 Chr.3: 192143992-192143992 on Build GRCh38
rs374897358 Chr.3: 192144027-192144027 on Build GRCh38
rs750508081 Chr.3: 192143999-192143999 on Build GRCh38
rs765046718 Chr.3: 192143994-192143994 on Build GRCh38
rs948132052 Chr.3: 192144030-192144030 on Build GRCh38
rs761890788 Chr.3: 192143991-192143991 on Build GRCh38
rs181770070 Chr.3: 192144139-192144139 on Build GRCh38
rs17852067 Chr.3: 192144109-192144109 on Build GRCh38
rs370723852 Chr.3: 192143988-192143988 on Build GRCh38
rs1015496615 Chr.3: 192144018-192144018 on Build GRCh38
rs991957244 Chr.3: 192144153-192144153 on Build GRCh38
rs769374802 Chr.3: 192144100-192144100 on Build GRCh38
rs771839343 Chr.3: 192144144-192144144 on Build GRCh38
rs758536451 Chr.3: 192144007-192144007 on Build GRCh38
rs759265723 Chr.3: 192143967-192143967 on Build GRCh38
rs775312046 Chr.3: 192144146-192144146 on Build GRCh38
rs939315978 Chr.3: 192143970-192143970 on Build GRCh38
rs774298359 Chr.3: 192143964-192143964 on Build GRCh38
rs770485100 Chr.3: 192144112-192144112 on Build GRCh38
rs780659108 Chr.3: 192144014-192144014 on Build GRCh38
rs3732883 Chr.3: 192144126-192144126 on Build GRCh38
rs752116921 Chr.3: 192144016-192144016 on Build GRCh38
rs368025154 Chr.3: 192144106-192144106 on Build GRCh38
rs369115025 Chr.3: 192144105-192144105 on Build GRCh38
rs774204331 Chr.3: 192144115-192144115 on Build GRCh38
rs374756136 Chr.3: 192143945-192143945 on Build GRCh38
rs190025961 Chr.3: 192144076-192144076 on Build GRCh38
rs781728534 Chr.3: 192144057-192144057 on Build GRCh38
rs765336878 Chr.3: 192143995-192143995 on Build GRCh38
rs767300269 Chr.3: 192143987-192143987 on Build GRCh38
rs200309674 Chr.3: 192144029-192144029 on Build GRCh38

View Full Product Details