Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 74986053-74986306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD1
Entrez Gene ID
83690
Gene Name
cysteine rich secretory protein LCCL domain containing 1
Gene Aliases
CRISP-10, CRISP10, LCRISP1
UniGene
Hs.436542
Gene Chromosome Location
Chr.8: 74984473-75034558 on Build GRCh38
Transcripts
NM_031461.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4476307 c.205C>T p.H69Y Chr.8: 74986192-74986192 on Build GRCh38
4509175 c.79C>T p.P27S Chr.8: 74986066-74986066 on Build GRCh38
6013191 c.120G>A p.M40I Chr.8: 74986107-74986107 on Build GRCh38
1202293 c.158G>T p.G53V Chr.8: 74986145-74986145 on Build GRCh38
3901537 c.217C>T p.R73* Chr.8: 74986204-74986204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747042310 Chr.8: 74986180-74986180 on Build GRCh38
rs765806338 Chr.8: 74986101-74986101 on Build GRCh38
rs969789453 Chr.8: 74986089-74986089 on Build GRCh38
rs779803912 Chr.8: 74986246-74986247 on Build GRCh38
rs376078635 Chr.8: 74986097-74986097 on Build GRCh38
rs779143565 Chr.8: 74986156-74986156 on Build GRCh38
rs1029490514 Chr.8: 74986083-74986083 on Build GRCh38
rs374697726 Chr.8: 74986177-74986177 on Build GRCh38
rs762504385 Chr.8: 74986246-74986246 on Build GRCh38
rs751576713 Chr.8: 74986114-74986114 on Build GRCh38
rs371100188 Chr.8: 74986222-74986222 on Build GRCh38
rs777138461 Chr.8: 74986085-74986085 on Build GRCh38
rs754925160 Chr.8: 74986122-74986122 on Build GRCh38
rs749150134 Chr.8: 74986254-74986255 on Build GRCh38
rs138941019 Chr.8: 74986143-74986143 on Build GRCh38
rs561721335 Chr.8: 74986158-74986158 on Build GRCh38
rs150274198 Chr.8: 74986088-74986088 on Build GRCh38
rs769320129 Chr.8: 74986188-74986188 on Build GRCh38
rs553885247 Chr.8: 74986254-74986254 on Build GRCh38
rs777230677 Chr.8: 74986190-74986190 on Build GRCh38
rs765061965 Chr.8: 74986251-74986251 on Build GRCh38
rs955216207 Chr.8: 74986084-74986084 on Build GRCh38
rs547219812 Chr.8: 74986220-74986220 on Build GRCh38
rs376754208 Chr.8: 74986165-74986165 on Build GRCh38
rs187003238 Chr.8: 74986253-74986253 on Build GRCh38
rs531674560 Chr.8: 74986148-74986148 on Build GRCh38
rs141929155 Chr.8: 74986080-74986080 on Build GRCh38
rs868463412 Chr.8: 74986073-74986073 on Build GRCh38
rs772987800 Chr.8: 74986227-74986227 on Build GRCh38
rs201928150 Chr.8: 74986095-74986095 on Build GRCh38
rs755877762 Chr.8: 74986151-74986151 on Build GRCh38
rs564224452 Chr.8: 74986137-74986137 on Build GRCh38
rs35476923 Chr.8: 74986244-74986245 on Build GRCh38
rs908306401 Chr.8: 74986175-74986175 on Build GRCh38
rs7821659 Chr.8: 74986266-74986266 on Build GRCh38
rs944868313 Chr.8: 74986231-74986231 on Build GRCh38
rs762231023 Chr.8: 74986086-74986086 on Build GRCh38
rs199940123 Chr.8: 74986090-74986090 on Build GRCh38
rs951013461 Chr.8: 74986189-74986189 on Build GRCh38

View Full Product Details