Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45786342-45786567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMWD
Entrez Gene ID
1762
Gene Name
dystrophia myotonica, WD repeat containing
Gene Aliases
D19S593E, DMR-N9, DMRN9, gene59
UniGene
Hs.515474
Gene Chromosome Location
Chr.19: 45783006-45792802 on Build GRCh38
Transcripts
NM_004943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712309 c.1152G>T p.E384D Chr.19: 45786344-45786344 on Build GRCh38
5074359 c.998_999insTG p.W334fs*10 Chr.19: 45786497-45786498 on Build GRCh38
6151466 c.987G>T p.L329L Chr.19: 45786509-45786509 on Build GRCh38
260449 c.939C>T p.F313F Chr.19: 45786557-45786557 on Build GRCh38
3535983 c.1052C>T p.T351I Chr.19: 45786444-45786444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147468425 Chr.19: 45786541-45786541 on Build GRCh38
rs374226582 Chr.19: 45786481-45786481 on Build GRCh38
rs779919971 Chr.19: 45786467-45786467 on Build GRCh38
rs767838131 Chr.19: 45786427-45786427 on Build GRCh38
rs757318714 Chr.19: 45786544-45786544 on Build GRCh38
rs764245481 Chr.19: 45786540-45786540 on Build GRCh38
rs1012841605 Chr.19: 45786419-45786419 on Build GRCh38
rs201586261 Chr.19: 45786485-45786485 on Build GRCh38
rs753023889 Chr.19: 45786533-45786533 on Build GRCh38
rs199578687 Chr.19: 45786443-45786443 on Build GRCh38
rs761530310 Chr.19: 45786380-45786380 on Build GRCh38
rs760894833 Chr.19: 45786536-45786536 on Build GRCh38
rs142998214 Chr.19: 45786461-45786461 on Build GRCh38
rs368040644 Chr.19: 45786402-45786402 on Build GRCh38
rs766736520 Chr.19: 45786413-45786413 on Build GRCh38
rs1026410121 Chr.19: 45786386-45786386 on Build GRCh38
rs770472820 Chr.19: 45786509-45786509 on Build GRCh38
rs772556490 Chr.19: 45786409-45786409 on Build GRCh38
rs756284542 Chr.19: 45786442-45786442 on Build GRCh38
rs372611310 Chr.19: 45786428-45786428 on Build GRCh38
rs774761042 Chr.19: 45786519-45786519 on Build GRCh38
rs763519027 Chr.19: 45786410-45786410 on Build GRCh38
rs767895573 Chr.19: 45786531-45786531 on Build GRCh38
rs752095405 Chr.19: 45786418-45786418 on Build GRCh38
rs749262615 Chr.19: 45786450-45786450 on Build GRCh38
rs762430824 Chr.19: 45786501-45786501 on Build GRCh38
rs545024316 Chr.19: 45786490-45786490 on Build GRCh38
rs760014095 Chr.19: 45786528-45786528 on Build GRCh38
rs1016685124 Chr.19: 45786434-45786434 on Build GRCh38
rs564825288 Chr.19: 45786547-45786547 on Build GRCh38
rs768617176 Chr.19: 45786476-45786476 on Build GRCh38
rs755365606 Chr.19: 45786421-45786421 on Build GRCh38
rs772790523 Chr.19: 45786492-45786492 on Build GRCh38
rs776459907 Chr.19: 45786365-45786365 on Build GRCh38
rs771751757 Chr.19: 45786517-45786518 on Build GRCh38
rs188331078 Chr.19: 45786486-45786486 on Build GRCh38
rs746931183 Chr.19: 45786470-45786470 on Build GRCh38
rs776514929 Chr.19: 45786477-45786477 on Build GRCh38

View Full Product Details