Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 180525-180766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBQ1
Entrez Gene ID
3049
Gene Name
hemoglobin subunit theta 1
Gene Aliases
HBQ
UniGene
Hs.247921
Gene Chromosome Location
Chr.16: 180334-181179 on Build GRCh38
Transcripts
NM_005331.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291736 c.86C>T p.A29V Chr.16: 180572-180572 on Build GRCh38
1209336 c.179G>A p.G60D Chr.16: 180749-180749 on Build GRCh38
5552449 c.193G>A p.D65N Chr.16: 180763-180763 on Build GRCh38
7370697 c.91G>A p.E31K Chr.16: 180577-180577 on Build GRCh38
4638944 c.67G>A p.G23S Chr.16: 180553-180553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760444390 Chr.16: 180631-180631 on Build GRCh38
rs775317264 Chr.16: 180748-180748 on Build GRCh38
rs780034510 Chr.16: 180592-180592 on Build GRCh38
rs771806135 Chr.16: 180628-180628 on Build GRCh38
rs1043992156 Chr.16: 180614-180614 on Build GRCh38
rs934399582 Chr.16: 180591-180591 on Build GRCh38
rs769113173 Chr.16: 180617-180617 on Build GRCh38
rs770545009 Chr.16: 180633-180633 on Build GRCh38
rs767682483 Chr.16: 180674-180674 on Build GRCh38
rs769721062 Chr.16: 180747-180747 on Build GRCh38
rs867998291 Chr.16: 180706-180706 on Build GRCh38
rs1018537253 Chr.16: 180637-180637 on Build GRCh38
rs539517809 Chr.16: 180690-180690 on Build GRCh38
rs779273965 Chr.16: 180618-180618 on Build GRCh38
rs369065332 Chr.16: 180736-180736 on Build GRCh38
rs749778892 Chr.16: 180611-180611 on Build GRCh38
rs529280017 Chr.16: 180564-180564 on Build GRCh38
rs147305378 Chr.16: 180734-180734 on Build GRCh38
rs939536278 Chr.16: 180556-180556 on Build GRCh38
rs991862699 Chr.16: 180744-180744 on Build GRCh38
rs781009419 Chr.16: 180730-180731 on Build GRCh38
rs776138470 Chr.16: 180738-180738 on Build GRCh38
rs1051306330 Chr.16: 180662-180662 on Build GRCh38
rs533978818 Chr.16: 180746-180746 on Build GRCh38
rs755458346 Chr.16: 180712-180712 on Build GRCh38
rs951470419 Chr.16: 180719-180719 on Build GRCh38
rs1042860586 Chr.16: 180578-180578 on Build GRCh38
rs11863726 Chr.16: 180579-180579 on Build GRCh38
rs776065252 Chr.16: 180634-180634 on Build GRCh38
rs79277966 Chr.16: 180644-180644 on Build GRCh38
rs772534431 Chr.16: 180732-180732 on Build GRCh38
rs773029849 Chr.16: 180630-180630 on Build GRCh38
rs781404561 Chr.16: 180555-180555 on Build GRCh38
rs946988830 Chr.16: 180586-180586 on Build GRCh38
rs746828470 Chr.16: 180585-180586 on Build GRCh38
rs995418856 Chr.16: 180726-180726 on Build GRCh38
rs1007598723 Chr.16: 180632-180632 on Build GRCh38
rs748479435 Chr.16: 180627-180627 on Build GRCh38
rs563261796 Chr.16: 180666-180666 on Build GRCh38
rs148010777 Chr.16: 180722-180722 on Build GRCh38
rs144518427 Chr.16: 180708-180708 on Build GRCh38
rs746648542 Chr.16: 180735-180735 on Build GRCh38
rs762758652 Chr.16: 180668-180668 on Build GRCh38
rs969798123 Chr.16: 180743-180743 on Build GRCh38
rs754757619 Chr.16: 180630-180630 on Build GRCh38
rs76613236 Chr.16: 180725-180725 on Build GRCh38
rs1004356061 Chr.16: 180694-180694 on Build GRCh38
rs995714897 Chr.16: 180669-180669 on Build GRCh38
rs181293989 Chr.16: 180640-180640 on Build GRCh38
rs753870983 Chr.16: 180711-180711 on Build GRCh38
rs756374513 Chr.16: 180698-180698 on Build GRCh38

View Full Product Details