Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 677647-677900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL3
Entrez Gene ID
10272
Gene Name
follistatin like 3
Gene Aliases
FLRG, FSRP
UniGene
Hs.529038
Gene Chromosome Location
Chr.19: 676389-683392 on Build GRCh38
Transcripts
NM_005860.2

SNP Detail

SNP ID Chromosome Location
rs565566272 Chr.19: 677881-677881 on Build GRCh38
rs768772221 Chr.19: 677710-677710 on Build GRCh38
rs762487681 Chr.19: 677866-677866 on Build GRCh38
rs941847025 Chr.19: 677693-677693 on Build GRCh38
rs1010613141 Chr.19: 677667-677667 on Build GRCh38
rs1043420151 Chr.19: 677864-677864 on Build GRCh38
rs768005904 Chr.19: 677868-677868 on Build GRCh38
rs376784869 Chr.19: 677776-677776 on Build GRCh38
rs767814124 Chr.19: 677697-677697 on Build GRCh38
rs755679092 Chr.19: 677865-677865 on Build GRCh38
rs138071677 Chr.19: 677875-677875 on Build GRCh38
rs766698556 Chr.19: 677882-677882 on Build GRCh38
rs563648903 Chr.19: 677696-677696 on Build GRCh38
rs776298851 Chr.19: 677858-677858 on Build GRCh38
rs916136442 Chr.19: 677862-677862 on Build GRCh38
rs142039044 Chr.19: 677880-677880 on Build GRCh38
rs755282319 Chr.19: 677788-677788 on Build GRCh38
rs1007149584 Chr.19: 677763-677763 on Build GRCh38
rs376007903 Chr.19: 677811-677811 on Build GRCh38
rs547270934 Chr.19: 677822-677822 on Build GRCh38
rs549110256 Chr.19: 677735-677735 on Build GRCh38
rs145464132 Chr.19: 677832-677832 on Build GRCh38
rs746769033 Chr.19: 677826-677826 on Build GRCh38
rs760521314 Chr.19: 677819-677821 on Build GRCh38
rs914870713 Chr.19: 677859-677859 on Build GRCh38
rs375138817 Chr.19: 677680-677680 on Build GRCh38
rs897773052 Chr.19: 677697-677697 on Build GRCh38
rs764664559 Chr.19: 677749-677749 on Build GRCh38
rs756567648 Chr.19: 677782-677782 on Build GRCh38
rs772104841 Chr.19: 677820-677820 on Build GRCh38
rs997430961 Chr.19: 677707-677707 on Build GRCh38
rs759145114 Chr.19: 677860-677860 on Build GRCh38
rs1034865279 Chr.19: 677706-677706 on Build GRCh38
rs115178410 Chr.19: 677732-677732 on Build GRCh38
rs375939246 Chr.19: 677797-677797 on Build GRCh38
rs752232730 Chr.19: 677767-677767 on Build GRCh38
rs993722508 Chr.19: 677716-677716 on Build GRCh38
rs529189263 Chr.19: 677787-677787 on Build GRCh38
rs762254801 Chr.19: 677772-677772 on Build GRCh38
rs769455629 Chr.19: 677861-677861 on Build GRCh38
rs923330405 Chr.19: 677824-677824 on Build GRCh38

View Full Product Details