Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99849942-99850179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMSS1
Entrez Gene ID
84319
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
C3orf26
UniGene
Hs.581320
Gene Chromosome Location
Chr.3: 99817834-100178632 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
UniGene
Hs.104672
Gene Chromosome Location
Chr.3: 99833144-100114513 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594140 c.1718C>T p.A573V Chr.3: 99849958-99849958 on Build GRCh38
7175987 c.1678G>C p.V560L Chr.3: 99849998-99849998 on Build GRCh38
7175988 c.1678G>C p.V560L Chr.3: 99849998-99849998 on Build GRCh38
1594138 c.1523G>T p.S508I Chr.3: 99850153-99850153 on Build GRCh38
5585228 c.1511G>A p.R504Q Chr.3: 99850165-99850165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760939603 Chr.3: 99850049-99850049 on Build GRCh38
rs376021359 Chr.3: 99850022-99850022 on Build GRCh38
rs754959682 Chr.3: 99850107-99850109 on Build GRCh38
rs374609337 Chr.3: 99850118-99850118 on Build GRCh38
rs760776400 Chr.3: 99849984-99849984 on Build GRCh38
rs922119415 Chr.3: 99850135-99850135 on Build GRCh38
rs763948053 Chr.3: 99850086-99850086 on Build GRCh38
rs777759195 Chr.3: 99849998-99849998 on Build GRCh38
rs558970547 Chr.3: 99850023-99850023 on Build GRCh38
rs769101256 Chr.3: 99850024-99850024 on Build GRCh38
rs751454474 Chr.3: 99850003-99850003 on Build GRCh38
rs980786696 Chr.3: 99849969-99849969 on Build GRCh38
rs377141445 Chr.3: 99850074-99850074 on Build GRCh38
rs367792130 Chr.3: 99849996-99849996 on Build GRCh38
rs966744961 Chr.3: 99850057-99850057 on Build GRCh38
rs750219567 Chr.3: 99850143-99850143 on Build GRCh38
rs766978391 Chr.3: 99850066-99850066 on Build GRCh38
rs750578542 Chr.3: 99849981-99849981 on Build GRCh38
rs766323387 Chr.3: 99849985-99849985 on Build GRCh38
rs187341961 Chr.3: 99850099-99850099 on Build GRCh38
rs780992749 Chr.3: 99850018-99850018 on Build GRCh38
rs577284397 Chr.3: 99850035-99850035 on Build GRCh38
rs930900564 Chr.3: 99850013-99850013 on Build GRCh38
rs369327422 Chr.3: 99850038-99850038 on Build GRCh38
rs901575559 Chr.3: 99850088-99850088 on Build GRCh38
rs201193835 Chr.3: 99850112-99850112 on Build GRCh38
rs769449136 Chr.3: 99850019-99850019 on Build GRCh38
rs374712984 Chr.3: 99849994-99849994 on Build GRCh38
rs998718397 Chr.3: 99850102-99850102 on Build GRCh38
rs867196616 Chr.3: 99849990-99849990 on Build GRCh38
rs373504477 Chr.3: 99850039-99850039 on Build GRCh38
rs559551588 Chr.3: 99850041-99850041 on Build GRCh38
rs563640451 Chr.3: 99850082-99850082 on Build GRCh38
rs751515338 Chr.3: 99850057-99850058 on Build GRCh38
rs780900196 Chr.3: 99850124-99850125 on Build GRCh38
rs371475352 Chr.3: 99850020-99850020 on Build GRCh38
rs139483111 Chr.3: 99850065-99850065 on Build GRCh38
rs757057504 Chr.3: 99850004-99850004 on Build GRCh38
rs767739889 Chr.3: 99849978-99849978 on Build GRCh38

View Full Product Details