Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53270591-53270836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPL1
Entrez Gene ID
9700
Gene Name
extra spindle pole bodies like 1, separase
Gene Aliases
ESP1, SEPA
UniGene
Hs.153479
Gene Chromosome Location
Chr.12: 53268257-53293643 on Build GRCh38
Transcripts
NM_012291.4 XM_006719705.3 XM_011539024.2 XM_011539025.2 XM_017020253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681698 c.1300G>A p.V434I Chr.12: 53270729-53270729 on Build GRCh38
3462636 c.1369+1G>A p.? Chr.12: 53270799-53270799 on Build GRCh38
5659915 c.1255G>C p.D419H Chr.12: 53270684-53270684 on Build GRCh38
191162 c.1349C>T p.T450M Chr.12: 53270778-53270778 on Build GRCh38
4445897 c.1249-3C>T p.? Chr.12: 53270675-53270675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201916454 Chr.12: 53270675-53270675 on Build GRCh38
rs1035862635 Chr.12: 53270749-53270749 on Build GRCh38
rs745360465 Chr.12: 53270663-53270663 on Build GRCh38
rs762702966 Chr.12: 53270636-53270639 on Build GRCh38
rs773546919 Chr.12: 53270720-53270720 on Build GRCh38
rs896703388 Chr.12: 53270617-53270617 on Build GRCh38
rs1005321855 Chr.12: 53270613-53270613 on Build GRCh38
rs751448597 Chr.12: 53270652-53270652 on Build GRCh38
rs141048890 Chr.12: 53270710-53270710 on Build GRCh38
rs750321552 Chr.12: 53270788-53270788 on Build GRCh38
rs771426538 Chr.12: 53270729-53270729 on Build GRCh38
rs181249188 Chr.12: 53270779-53270779 on Build GRCh38
rs777571325 Chr.12: 53270645-53270645 on Build GRCh38
rs755593883 Chr.12: 53270671-53270671 on Build GRCh38
rs149663100 Chr.12: 53270728-53270728 on Build GRCh38
rs371969495 Chr.12: 53270763-53270763 on Build GRCh38
rs371851364 Chr.12: 53270659-53270659 on Build GRCh38
rs144769405 Chr.12: 53270787-53270787 on Build GRCh38
rs756038295 Chr.12: 53270797-53270797 on Build GRCh38
rs759962350 Chr.12: 53270747-53270747 on Build GRCh38
rs367983712 Chr.12: 53270650-53270650 on Build GRCh38
rs765316711 Chr.12: 53270758-53270758 on Build GRCh38
rs1012952522 Chr.12: 53270626-53270626 on Build GRCh38
rs1000603702 Chr.12: 53270680-53270680 on Build GRCh38
rs373722072 Chr.12: 53270641-53270641 on Build GRCh38
rs779435119 Chr.12: 53270702-53270702 on Build GRCh38
rs779567538 Chr.12: 53270798-53270798 on Build GRCh38
rs1022768813 Chr.12: 53270679-53270679 on Build GRCh38
rs375342185 Chr.12: 53270806-53270806 on Build GRCh38
rs777062092 Chr.12: 53270744-53270744 on Build GRCh38
rs1056565680 Chr.12: 53270616-53270616 on Build GRCh38
rs751393542 Chr.12: 53270769-53270769 on Build GRCh38
rs758576383 Chr.12: 53270634-53270634 on Build GRCh38
rs754631652 Chr.12: 53270811-53270811 on Build GRCh38
rs1023293014 Chr.12: 53270646-53270646 on Build GRCh38
rs781343942 Chr.12: 53270656-53270656 on Build GRCh38
rs748930510 Chr.12: 53270707-53270707 on Build GRCh38

View Full Product Details