Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84838399-84838672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD2
Entrez Gene ID
83716
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Gene Aliases
CRISP11, LCRISP2
UniGene
Hs.513779
Gene Chromosome Location
Chr.16: 84819981-84909510 on Build GRCh38
Transcripts
NM_031476.3 XM_005256190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7098411 c.77C>G p.P26R Chr.16: 84838572-84838572 on Build GRCh38
6635562 c.177C>T p.I59I Chr.16: 84838672-84838672 on Build GRCh38
4530798 c.172G>A p.E58K Chr.16: 84838667-84838667 on Build GRCh38
7152525 c.24C>A p.V8V Chr.16: 84838519-84838519 on Build GRCh38
3387641 c.168G>A p.K56K Chr.16: 84838663-84838663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746726440 Chr.16: 84838627-84838627 on Build GRCh38
rs141769268 Chr.16: 84838572-84838572 on Build GRCh38
rs375349489 Chr.16: 84838488-84838488 on Build GRCh38
rs760870426 Chr.16: 84838609-84838609 on Build GRCh38
rs756976159 Chr.16: 84838630-84838630 on Build GRCh38
rs767752138 Chr.16: 84838502-84838502 on Build GRCh38
rs201151162 Chr.16: 84838633-84838633 on Build GRCh38
rs146258520 Chr.16: 84838619-84838619 on Build GRCh38
rs970980736 Chr.16: 84838455-84838455 on Build GRCh38
rs770252855 Chr.16: 84838470-84838470 on Build GRCh38
rs369116191 Chr.16: 84838474-84838474 on Build GRCh38
rs766136257 Chr.16: 84838540-84838540 on Build GRCh38
rs570628920 Chr.16: 84838620-84838620 on Build GRCh38
rs774511408 Chr.16: 84838611-84838611 on Build GRCh38
rs978210991 Chr.16: 84838459-84838459 on Build GRCh38
rs978308294 Chr.16: 84838420-84838420 on Build GRCh38
rs760335963 Chr.16: 84838531-84838531 on Build GRCh38
rs766010932 Chr.16: 84838532-84838532 on Build GRCh38
rs1036189956 Chr.16: 84838608-84838608 on Build GRCh38
rs755939119 Chr.16: 84838529-84838529 on Build GRCh38
rs147872508 Chr.16: 84838560-84838560 on Build GRCh38
rs373176982 Chr.16: 84838638-84838638 on Build GRCh38
rs746638399 Chr.16: 84838570-84838570 on Build GRCh38
rs760000874 Chr.16: 84838464-84838464 on Build GRCh38
rs996045027 Chr.16: 84838535-84838535 on Build GRCh38
rs372585965 Chr.16: 84838463-84838463 on Build GRCh38
rs747623376 Chr.16: 84838552-84838552 on Build GRCh38
rs753868566 Chr.16: 84838543-84838543 on Build GRCh38
rs753577527 Chr.16: 84838618-84838618 on Build GRCh38
rs867693538 Chr.16: 84838629-84838629 on Build GRCh38
rs750608727 Chr.16: 84838505-84838505 on Build GRCh38
rs148905802 Chr.16: 84838650-84838650 on Build GRCh38
rs765107752 Chr.16: 84838621-84838621 on Build GRCh38
rs761897209 Chr.16: 84838501-84838501 on Build GRCh38
rs1546124 Chr.16: 84838445-84838445 on Build GRCh38
rs914614405 Chr.16: 84838428-84838428 on Build GRCh38
rs147841355 Chr.16: 84838601-84838601 on Build GRCh38
rs775994375 Chr.16: 84838575-84838575 on Build GRCh38
rs200402899 Chr.16: 84838480-84838480 on Build GRCh38
rs554885433 Chr.16: 84838431-84838431 on Build GRCh38
rs192527000 Chr.16: 84838649-84838649 on Build GRCh38
rs207476236 Chr.16: 84838537-84838537 on Build GRCh38
rs757875800 Chr.16: 84838553-84838553 on Build GRCh38
rs764303714 Chr.16: 84838479-84838479 on Build GRCh38
rs778841679 Chr.16: 84838550-84838550 on Build GRCh38
rs139901174 Chr.16: 84838576-84838576 on Build GRCh38
rs200281033 Chr.16: 84838637-84838637 on Build GRCh38
rs541302905 Chr.16: 84838577-84838577 on Build GRCh38
rs759240279 Chr.16: 84838629-84838630 on Build GRCh38
rs143123171 Chr.16: 84838452-84838452 on Build GRCh38
rs747899661 Chr.16: 84838462-84838462 on Build GRCh38
rs763357822 Chr.16: 84838478-84838478 on Build GRCh38
rs777408980 Chr.16: 84838625-84838625 on Build GRCh38
rs569603671 Chr.16: 84838634-84838634 on Build GRCh38
rs965984074 Chr.16: 84838589-84838589 on Build GRCh38
rs201798041 Chr.16: 84838623-84838623 on Build GRCh38
rs148237290 Chr.16: 84838549-84838549 on Build GRCh38
rs145368506 Chr.16: 84838597-84838597 on Build GRCh38
rs780937131 Chr.16: 84838632-84838632 on Build GRCh38
rs938929677 Chr.16: 84838512-84838512 on Build GRCh38
rs987184821 Chr.16: 84838585-84838585 on Build GRCh38

View Full Product Details