Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171553139-171553380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBRD1
Entrez Gene ID
79901
Gene Name
cytochrome b reductase 1
Gene Aliases
CYB561A2, DCYTB, FRRS3
UniGene
Hs.221941
Gene Chromosome Location
Chr.2: 171522247-171558133 on Build GRCh38
Transcripts
NM_001127383.1 NM_001256909.1 NM_024843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603333 c.434C>T p.P145L Chr.2: 171553377-171553377 on Build GRCh38
5394950 c.427C>T p.L143L Chr.2: 171553370-171553370 on Build GRCh38
5439135 c.423C>G p.V141V Chr.2: 171553366-171553366 on Build GRCh38
3895350 c.406C>T p.L136F Chr.2: 171553349-171553349 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958525533 Chr.2: 171553264-171553264 on Build GRCh38
rs569201460 Chr.2: 171553297-171553297 on Build GRCh38
rs958771066 Chr.2: 171553326-171553326 on Build GRCh38
rs2542941 Chr.2: 171553211-171553211 on Build GRCh38
rs996877376 Chr.2: 171553208-171553208 on Build GRCh38
rs779671593 Chr.2: 171553318-171553318 on Build GRCh38
rs751203760 Chr.2: 171553213-171553213 on Build GRCh38
rs776489350 Chr.2: 171553331-171553331 on Build GRCh38
rs1040480698 Chr.2: 171553183-171553183 on Build GRCh38
rs868251308 Chr.2: 171553252-171553252 on Build GRCh38
rs768596361 Chr.2: 171553222-171553222 on Build GRCh38
rs1034175461 Chr.2: 171553257-171553257 on Build GRCh38
rs767386737 Chr.2: 171553306-171553311 on Build GRCh38
rs532659923 Chr.2: 171553232-171553232 on Build GRCh38
rs776678594 Chr.2: 171553187-171553188 on Build GRCh38
rs536929544 Chr.2: 171553323-171553323 on Build GRCh38
rs1024010524 Chr.2: 171553336-171553336 on Build GRCh38
rs142443896 Chr.2: 171553205-171553205 on Build GRCh38
rs143524630 Chr.2: 171553281-171553281 on Build GRCh38
rs760421926 Chr.2: 171553332-171553332 on Build GRCh38
rs746965513 Chr.2: 171553322-171553322 on Build GRCh38
rs775328160 Chr.2: 171553307-171553311 on Build GRCh38
rs1004078667 Chr.2: 171553269-171553269 on Build GRCh38
rs550982499 Chr.2: 171553233-171553233 on Build GRCh38
rs888388282 Chr.2: 171553219-171553219 on Build GRCh38

View Full Product Details