Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7727067-7727338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
UniGene
Hs.367649
Gene Chromosome Location
Chr.17: 7717354-7833744 on Build GRCh38
Transcripts
NM_001303270.1 NM_020877.3 XM_011523663.1 XM_011523664.2 XM_011523666.1 XM_011523667.2 XM_011523670.2 XM_017024219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6250832 c.281G>T p.W94L Chr.17: 7727174-7727174 on Build GRCh38
6841759 c.399G>T p.Q133H Chr.17: 7727292-7727292 on Build GRCh38
5343038 c.399+5G>A p.? Chr.17: 7727297-7727297 on Build GRCh38
5343037 c.399+5G>A p.? Chr.17: 7727297-7727297 on Build GRCh38
6016469 c.399+6T>A p.? Chr.17: 7727298-7727298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776571109 Chr.17: 7727277-7727277 on Build GRCh38
rs113638848 Chr.17: 7727205-7727205 on Build GRCh38
rs556580433 Chr.17: 7727162-7727162 on Build GRCh38
rs369384877 Chr.17: 7727126-7727126 on Build GRCh38
rs373888338 Chr.17: 7727141-7727141 on Build GRCh38
rs367657705 Chr.17: 7727274-7727274 on Build GRCh38
rs189225191 Chr.17: 7727295-7727295 on Build GRCh38
rs754564026 Chr.17: 7727207-7727207 on Build GRCh38
rs747731188 Chr.17: 7727140-7727140 on Build GRCh38
rs374375369 Chr.17: 7727168-7727168 on Build GRCh38
rs762100967 Chr.17: 7727286-7727286 on Build GRCh38
rs931490511 Chr.17: 7727268-7727268 on Build GRCh38
rs981482963 Chr.17: 7727090-7727090 on Build GRCh38
rs35406368 Chr.17: 7727148-7727148 on Build GRCh38
rs377077879 Chr.17: 7727147-7727147 on Build GRCh38
rs201592016 Chr.17: 7727181-7727181 on Build GRCh38
rs755653472 Chr.17: 7727222-7727222 on Build GRCh38
rs749038332 Chr.17: 7727239-7727239 on Build GRCh38
rs371827721 Chr.17: 7727297-7727297 on Build GRCh38
rs74527020 Chr.17: 7727271-7727271 on Build GRCh38
rs758904095 Chr.17: 7727110-7727110 on Build GRCh38
rs763346780 Chr.17: 7727296-7727296 on Build GRCh38
rs199656868 Chr.17: 7727115-7727115 on Build GRCh38
rs775730744 Chr.17: 7727163-7727163 on Build GRCh38
rs746465716 Chr.17: 7727122-7727122 on Build GRCh38
rs750998765 Chr.17: 7727108-7727108 on Build GRCh38
rs780553400 Chr.17: 7727213-7727213 on Build GRCh38
rs747899399 Chr.17: 7727221-7727221 on Build GRCh38
rs745780991 Chr.17: 7727250-7727250 on Build GRCh38
rs11867551 Chr.17: 7727187-7727187 on Build GRCh38
rs752030328 Chr.17: 7727204-7727204 on Build GRCh38
rs752116524 Chr.17: 7727315-7727315 on Build GRCh38
rs961323906 Chr.17: 7727119-7727119 on Build GRCh38
rs773589675 Chr.17: 7727292-7727292 on Build GRCh38
rs764054226 Chr.17: 7727169-7727169 on Build GRCh38
rs766823892 Chr.17: 7727199-7727199 on Build GRCh38
rs779127526 Chr.17: 7727106-7727106 on Build GRCh38
rs758920299 Chr.17: 7727196-7727196 on Build GRCh38
rs561549500 Chr.17: 7727288-7727288 on Build GRCh38
rs777632922 Chr.17: 7727227-7727227 on Build GRCh38
rs200612151 Chr.17: 7727183-7727183 on Build GRCh38
rs35664870 Chr.17: 7727192-7727192 on Build GRCh38
rs375449200 Chr.17: 7727317-7727317 on Build GRCh38
rs142363026 Chr.17: 7727246-7727246 on Build GRCh38

View Full Product Details