Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41292370-41292607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534380 c.833C>T p.S278F Chr.19: 41292420-41292420 on Build GRCh38
6681453 c.1132G>A p.A378T Chr.19: 41292377-41292377 on Build GRCh38
5981543 c.884T>C p.I295T Chr.19: 41292471-41292471 on Build GRCh38
6681447 c.1223G>A p.R408H Chr.19: 41292468-41292468 on Build GRCh38
4283349 c.882T>C p.R294R Chr.19: 41292469-41292469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763500949 Chr.19: 41292566-41292567 on Build GRCh38
rs58076789 Chr.19: 41292585-41292586 on Build GRCh38
rs779949595 Chr.19: 41292487-41292487 on Build GRCh38
rs58242393 Chr.19: 41292586-41292587 on Build GRCh38
rs57173622 Chr.19: 41292583-41292586 on Build GRCh38
rs200756551 Chr.19: 41292584-41292587 on Build GRCh38
rs112213182 Chr.19: 41292512-41292512 on Build GRCh38
rs867126206 Chr.19: 41292457-41292457 on Build GRCh38
rs528087616 Chr.19: 41292394-41292394 on Build GRCh38
rs1027943255 Chr.19: 41292587-41292587 on Build GRCh38
rs760978783 Chr.19: 41292557-41292561 on Build GRCh38
rs755713629 Chr.19: 41292433-41292433 on Build GRCh38
rs766697507 Chr.19: 41292417-41292417 on Build GRCh38
rs758463831 Chr.19: 41292482-41292482 on Build GRCh38
rs1009402429 Chr.19: 41292572-41292572 on Build GRCh38
rs763580084 Chr.19: 41292414-41292414 on Build GRCh38
rs746834749 Chr.19: 41292518-41292518 on Build GRCh38
rs66479074 Chr.19: 41292564-41292565 on Build GRCh38
rs756710629 Chr.19: 41292471-41292471 on Build GRCh38
rs778553397 Chr.19: 41292473-41292473 on Build GRCh38
rs769755884 Chr.19: 41292539-41292539 on Build GRCh38
rs112745840 Chr.19: 41292564-41292567 on Build GRCh38
rs1017666341 Chr.19: 41292573-41292573 on Build GRCh38
rs12974243 Chr.19: 41292407-41292407 on Build GRCh38
rs546143838 Chr.19: 41292561-41292561 on Build GRCh38
rs34099094 Chr.19: 41292587-41292588 on Build GRCh38
rs768935854 Chr.19: 41292525-41292525 on Build GRCh38
rs779518064 Chr.19: 41292564-41292569 on Build GRCh38
rs763871796 Chr.19: 41292481-41292481 on Build GRCh38
rs765715604 Chr.19: 41292409-41292409 on Build GRCh38
rs777003728 Chr.19: 41292532-41292532 on Build GRCh38
rs528919566 Chr.19: 41292576-41292576 on Build GRCh38
rs548291000 Chr.19: 41292567-41292568 on Build GRCh38
rs545165335 Chr.19: 41292410-41292410 on Build GRCh38
rs561465996 Chr.19: 41292568-41292568 on Build GRCh38
rs369738571 Chr.19: 41292533-41292533 on Build GRCh38
rs181859281 Chr.19: 41292585-41292585 on Build GRCh38
rs755396135 Chr.19: 41292563-41292564 on Build GRCh38
rs751968794 Chr.19: 41292425-41292425 on Build GRCh38
rs900526650 Chr.19: 41292578-41292578 on Build GRCh38
rs750740675 Chr.19: 41292536-41292537 on Build GRCh38
rs756764728 Chr.19: 41292584-41292585 on Build GRCh38
rs763764872 Chr.19: 41292455-41292455 on Build GRCh38
rs753332324 Chr.19: 41292467-41292467 on Build GRCh38

View Full Product Details