Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55871014-55871275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC1
Entrez Gene ID
55763
Gene Name
exocyst complex component 1
Gene Aliases
BM-102, SEC3, SEC3L1, SEC3P
UniGene
Hs.269665
Gene Chromosome Location
Chr.4: 55853616-55905078 on Build GRCh38
Transcripts
NM_001024924.1 NM_018261.3 NM_178237.2 XM_005265747.2 XM_005265748.1 XM_005265750.2 XM_017008405.1 XM_017008406.1 XM_017008407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633726 c.879A>C p.G293G Chr.4: 55871148-55871148 on Build GRCh38
734111 c.832-2A>G p.? Chr.4: 55871099-55871099 on Build GRCh38
6011476 c.849C>T p.A283A Chr.4: 55871118-55871118 on Build GRCh38
6100247 c.838A>G p.M280V Chr.4: 55871107-55871107 on Build GRCh38
4585113 c.909C>G p.A303A Chr.4: 55871178-55871178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748184834 Chr.4: 55871081-55871081 on Build GRCh38
rs1018429131 Chr.4: 55871071-55871071 on Build GRCh38
rs371142509 Chr.4: 55871175-55871175 on Build GRCh38
rs755252003 Chr.4: 55871194-55871194 on Build GRCh38
rs769623660 Chr.4: 55871093-55871093 on Build GRCh38
rs779073924 Chr.4: 55871200-55871200 on Build GRCh38
rs956968903 Chr.4: 55871110-55871110 on Build GRCh38
rs976966152 Chr.4: 55871078-55871078 on Build GRCh38
rs778979573 Chr.4: 55871079-55871079 on Build GRCh38
rs368504673 Chr.4: 55871108-55871108 on Build GRCh38
rs775075850 Chr.4: 55871096-55871096 on Build GRCh38
rs988128039 Chr.4: 55871134-55871134 on Build GRCh38
rs201913166 Chr.4: 55871154-55871154 on Build GRCh38
rs964220915 Chr.4: 55871073-55871073 on Build GRCh38
rs749522431 Chr.4: 55871059-55871059 on Build GRCh38
rs756502973 Chr.4: 55871186-55871186 on Build GRCh38
rs759306304 Chr.4: 55871092-55871092 on Build GRCh38
rs559479724 Chr.4: 55871166-55871166 on Build GRCh38
rs780472170 Chr.4: 55871187-55871187 on Build GRCh38
rs772125627 Chr.4: 55871216-55871216 on Build GRCh38
rs751148319 Chr.4: 55871158-55871158 on Build GRCh38
rs143045485 Chr.4: 55871178-55871178 on Build GRCh38
rs866068231 Chr.4: 55871183-55871183 on Build GRCh38
rs781172342 Chr.4: 55871220-55871220 on Build GRCh38
rs754095123 Chr.4: 55871192-55871192 on Build GRCh38
rs775374223 Chr.4: 55871236-55871236 on Build GRCh38
rs745819122 Chr.4: 55871227-55871227 on Build GRCh38
rs370540468 Chr.4: 55871090-55871090 on Build GRCh38
rs763787664 Chr.4: 55871143-55871143 on Build GRCh38
rs749724076 Chr.4: 55871245-55871248 on Build GRCh38
rs769557546 Chr.4: 55871231-55871231 on Build GRCh38
rs748274634 Chr.4: 55871207-55871207 on Build GRCh38
rs768696262 Chr.4: 55871075-55871075 on Build GRCh38
rs770893219 Chr.4: 55871088-55871088 on Build GRCh38
rs995497930 Chr.4: 55871062-55871062 on Build GRCh38

View Full Product Details