Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46940430-46940695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOSR2
Entrez Gene ID
9570
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
Bos1, EPM6, GS27
UniGene
Hs.463278
Gene Chromosome Location
Chr.17: 46923086-46975519 on Build GRCh38
Transcripts
NM_001321133.1 NM_001321134.1 NM_004287.4 NM_054022.3 XM_005257844.3 XM_006722190.3 XM_011525501.2 XM_011525502.2 XM_017025378.1 XM_017025379.1 XM_017025380.1 XM_017025381.1 XM_017025382.1 XM_017025383.1 XM_017025384.1 XM_017025385.1 XM_017025386.1 XM_017025387.1 XM_017025388.1 XM_017025389.1 XM_017025390.1 XM_017025391.1 XM_017025392.1 XM_017025393.1 XM_017025394.1 XM_017025395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4925535 c.584G>T p.G195V Chr.17: 46940555-46940555 on Build GRCh38
5386535 c.613delT p.F205fs*>9 Chr.17: 46940584-46940584 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747267756 Chr.17: 46940539-46940539 on Build GRCh38
rs374383310 Chr.17: 46940528-46940528 on Build GRCh38
rs749239417 Chr.17: 46940586-46940586 on Build GRCh38
rs369557972 Chr.17: 46940532-46940532 on Build GRCh38
rs889988218 Chr.17: 46940483-46940483 on Build GRCh38
rs969218389 Chr.17: 46940654-46940654 on Build GRCh38
rs148511622 Chr.17: 46940512-46940512 on Build GRCh38
rs11657819 Chr.17: 46940471-46940471 on Build GRCh38
rs140685046 Chr.17: 46940560-46940560 on Build GRCh38
rs745773590 Chr.17: 46940451-46940451 on Build GRCh38
rs758885350 Chr.17: 46940533-46940533 on Build GRCh38
rs1019401230 Chr.17: 46940541-46940541 on Build GRCh38
rs767466315 Chr.17: 46940597-46940597 on Build GRCh38
rs774352164 Chr.17: 46940595-46940595 on Build GRCh38
rs765974207 Chr.17: 46940605-46940605 on Build GRCh38
rs766295758 Chr.17: 46940546-46940547 on Build GRCh38
rs751561082 Chr.17: 46940553-46940553 on Build GRCh38
rs941354762 Chr.17: 46940600-46940600 on Build GRCh38
rs369846922 Chr.17: 46940671-46940671 on Build GRCh38
rs760466734 Chr.17: 46940601-46940601 on Build GRCh38
rs1005916066 Chr.17: 46940629-46940629 on Build GRCh38
rs746138669 Chr.17: 46940568-46940568 on Build GRCh38
rs769834243 Chr.17: 46940580-46940580 on Build GRCh38
rs755660289 Chr.17: 46940530-46940530 on Build GRCh38
rs757598206 Chr.17: 46940660-46940660 on Build GRCh38
rs778355377 Chr.17: 46940536-46940536 on Build GRCh38
rs147434599 Chr.17: 46940555-46940555 on Build GRCh38
rs142261376 Chr.17: 46940583-46940583 on Build GRCh38
rs575057184 Chr.17: 46940599-46940599 on Build GRCh38
rs757651569 Chr.17: 46940549-46940549 on Build GRCh38
rs758598475 Chr.17: 46940472-46940472 on Build GRCh38
rs753229227 Chr.17: 46940609-46940609 on Build GRCh38
rs767214928 Chr.17: 46940525-46940525 on Build GRCh38
rs377308635 Chr.17: 46940531-46940531 on Build GRCh38
rs764465941 Chr.17: 46940650-46940650 on Build GRCh38
rs758392 Chr.17: 46940493-46940493 on Build GRCh38
rs746086938 Chr.17: 46940664-46940664 on Build GRCh38
rs373654893 Chr.17: 46940661-46940661 on Build GRCh38
rs756390829 Chr.17: 46940665-46940665 on Build GRCh38
rs752132148 Chr.17: 46940651-46940651 on Build GRCh38
rs768422581 Chr.17: 46940594-46940594 on Build GRCh38
rs144662292 Chr.17: 46940463-46940463 on Build GRCh38
rs754519349 Chr.17: 46940633-46940633 on Build GRCh38

View Full Product Details