Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168042114-168042351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF25
Entrez Gene ID
3834
Gene Name
kinesin family member 25
Gene Aliases
KNSL3
UniGene
Hs.150013
Gene Chromosome Location
Chr.6: 168002543-168045106 on Build GRCh38
Transcripts
NM_005355.3 NM_030615.2 XM_011535802.2 XM_011535803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4559596 c.810G>A p.S270S Chr.6: 168042132-168042132 on Build GRCh38
402834 c.820G>T p.E274* Chr.6: 168042142-168042142 on Build GRCh38
396186 c.802G>T p.V268L Chr.6: 168042124-168042124 on Build GRCh38
5404803 c.820G>A p.E274K Chr.6: 168042142-168042142 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs977998113 Chr.6: 168042232-168042232 on Build GRCh38
rs546525129 Chr.6: 168042306-168042306 on Build GRCh38
rs936345600 Chr.6: 168042246-168042246 on Build GRCh38
rs750565768 Chr.6: 168042136-168042136 on Build GRCh38
rs943563819 Chr.6: 168042289-168042289 on Build GRCh38
rs117190346 Chr.6: 168042291-168042291 on Build GRCh38
rs545611039 Chr.6: 168042251-168042251 on Build GRCh38
rs758456030 Chr.6: 168042158-168042158 on Build GRCh38
rs970644404 Chr.6: 168042220-168042220 on Build GRCh38
rs990641173 Chr.6: 168042183-168042183 on Build GRCh38
rs201046260 Chr.6: 168042156-168042156 on Build GRCh38
rs140465870 Chr.6: 168042324-168042324 on Build GRCh38
rs998073562 Chr.6: 168042160-168042160 on Build GRCh38
rs138836160 Chr.6: 168042141-168042141 on Build GRCh38
rs574381631 Chr.6: 168042157-168042157 on Build GRCh38
rs746548312 Chr.6: 168042166-168042166 on Build GRCh38
rs929664424 Chr.6: 168042318-168042318 on Build GRCh38
rs910957424 Chr.6: 168042275-168042275 on Build GRCh38
rs140413393 Chr.6: 168042165-168042165 on Build GRCh38
rs56168144 Chr.6: 168042328-168042328 on Build GRCh38
rs151246487 Chr.6: 168042250-168042250 on Build GRCh38
rs146056201 Chr.6: 168042138-168042138 on Build GRCh38
rs756777726 Chr.6: 168042193-168042193 on Build GRCh38
rs923604082 Chr.6: 168042237-168042237 on Build GRCh38
rs761853452 Chr.6: 168042132-168042132 on Build GRCh38
rs143536760 Chr.6: 168042203-168042203 on Build GRCh38
rs776390430 Chr.6: 168042160-168042161 on Build GRCh38
rs61326012 Chr.6: 168042299-168042299 on Build GRCh38
rs998185226 Chr.6: 168042323-168042323 on Build GRCh38
rs752520655 Chr.6: 168042143-168042143 on Build GRCh38
rs1006216391 Chr.6: 168042331-168042331 on Build GRCh38
rs540998005 Chr.6: 168042135-168042135 on Build GRCh38
rs759221584 Chr.6: 168042181-168042201 on Build GRCh38
rs1033091496 Chr.6: 168042236-168042236 on Build GRCh38
rs867328122 Chr.6: 168042310-168042310 on Build GRCh38

View Full Product Details