Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16840391-16840616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUBN
Entrez Gene ID
8029
Gene Name
cubilin
Gene Aliases
IFCR, MGA1, gp280
UniGene
Hs.166206
Gene Chromosome Location
Chr.10: 16823966-17130492 on Build GRCh38
Transcripts
NM_001081.3 XM_011519709.2 XM_011519710.2 XM_011519711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4514753 c.9875C>T p.S3292L Chr.10: 16840487-16840487 on Build GRCh38
6206990 c.9931G>A p.D3311N Chr.10: 16840431-16840431 on Build GRCh38
6394832 c.9833G>T p.C3278F Chr.10: 16840529-16840529 on Build GRCh38
2131578 c.9911C>T p.S3304F Chr.10: 16840451-16840451 on Build GRCh38
397396 c.9964G>A p.V3322M Chr.10: 16840398-16840398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371665308 Chr.10: 16840534-16840534 on Build GRCh38
rs751837203 Chr.10: 16840528-16840528 on Build GRCh38
rs1001740365 Chr.10: 16840434-16840434 on Build GRCh38
rs760142969 Chr.10: 16840511-16840511 on Build GRCh38
rs752843169 Chr.10: 16840440-16840440 on Build GRCh38
rs527327584 Chr.10: 16840486-16840486 on Build GRCh38
rs756115968 Chr.10: 16840572-16840572 on Build GRCh38
rs757426720 Chr.10: 16840463-16840463 on Build GRCh38
rs761252412 Chr.10: 16840497-16840497 on Build GRCh38
rs911024813 Chr.10: 16840544-16840544 on Build GRCh38
rs745848165 Chr.10: 16840466-16840466 on Build GRCh38
rs143405367 Chr.10: 16840465-16840465 on Build GRCh38
rs775402701 Chr.10: 16840481-16840481 on Build GRCh38
rs375963302 Chr.10: 16840420-16840420 on Build GRCh38
rs143027674 Chr.10: 16840447-16840447 on Build GRCh38
rs754351697 Chr.10: 16840427-16840427 on Build GRCh38
rs769425968 Chr.10: 16840476-16840476 on Build GRCh38
rs765660437 Chr.10: 16840437-16840437 on Build GRCh38
rs529911077 Chr.10: 16840421-16840421 on Build GRCh38
rs777078292 Chr.10: 16840508-16840508 on Build GRCh38
rs566296480 Chr.10: 16840484-16840484 on Build GRCh38
rs181084844 Chr.10: 16840583-16840583 on Build GRCh38
rs747145299 Chr.10: 16840459-16840459 on Build GRCh38
rs763488541 Chr.10: 16840412-16840412 on Build GRCh38
rs369895669 Chr.10: 16840485-16840485 on Build GRCh38
rs777997194 Chr.10: 16840457-16840457 on Build GRCh38
rs757373345 Chr.10: 16840555-16840555 on Build GRCh38
rs202234356 Chr.10: 16840435-16840435 on Build GRCh38
rs750304590 Chr.10: 16840565-16840565 on Build GRCh38
rs376882010 Chr.10: 16840507-16840507 on Build GRCh38
rs758538084 Chr.10: 16840520-16840520 on Build GRCh38
rs703064 Chr.10: 16840519-16840519 on Build GRCh38
rs78729515 Chr.10: 16840562-16840562 on Build GRCh38
rs765490955 Chr.10: 16840514-16840514 on Build GRCh38
rs570241713 Chr.10: 16840521-16840521 on Build GRCh38

View Full Product Details