Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153790409-153790668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH3G
Entrez Gene ID
3421
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
Gene Aliases
H-IDHG
UniGene
Hs.410197
Gene Chromosome Location
Chr.X: 153785766-153794523 on Build GRCh38
Transcripts
NM_004135.3 NM_174869.2

SNP Detail

SNP ID Chromosome Location
rs782723361 Chr.X: 153790568-153790568 on Build GRCh38
rs782073740 Chr.X: 153790484-153790484 on Build GRCh38
rs782004123 Chr.X: 153790550-153790550 on Build GRCh38
rs782203483 Chr.X: 153790543-153790543 on Build GRCh38
rs997329137 Chr.X: 153790474-153790474 on Build GRCh38
rs782749368 Chr.X: 153790528-153790529 on Build GRCh38
rs782587933 Chr.X: 153790536-153790536 on Build GRCh38
rs782297401 Chr.X: 153790516-153790516 on Build GRCh38
rs782305868 Chr.X: 153790529-153790529 on Build GRCh38
rs374030294 Chr.X: 153790552-153790552 on Build GRCh38
rs782754939 Chr.X: 153790511-153790511 on Build GRCh38
rs781886066 Chr.X: 153790611-153790611 on Build GRCh38
rs782789740 Chr.X: 153790597-153790597 on Build GRCh38
rs782518845 Chr.X: 153790514-153790514 on Build GRCh38
rs371348645 Chr.X: 153790623-153790623 on Build GRCh38
rs781929270 Chr.X: 153790565-153790565 on Build GRCh38
rs369805805 Chr.X: 153790513-153790513 on Build GRCh38
rs782352933 Chr.X: 153790547-153790547 on Build GRCh38
rs782068621 Chr.X: 153790567-153790567 on Build GRCh38
rs782088157 Chr.X: 153790528-153790528 on Build GRCh38
rs781985688 Chr.X: 153790561-153790562 on Build GRCh38
rs377743175 Chr.X: 153790554-153790554 on Build GRCh38
rs781985265 Chr.X: 153790439-153790439 on Build GRCh38
rs781972713 Chr.X: 153790571-153790571 on Build GRCh38
rs782115500 Chr.X: 153790589-153790589 on Build GRCh38

View Full Product Details