Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123557416-123557671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G3
Entrez Gene ID
1456
Gene Name
casein kinase 1 gamma 3
Gene Aliases
CKI-gamma 3, CSNK1G3L
UniGene
Hs.129206
Gene Chromosome Location
Chr.5: 123512099-123617045 on Build GRCh38
Transcripts
NM_001031812.3 NM_001044723.2 NM_001270572.1 NM_001270573.1 NM_001270574.1 NM_004384.4 XM_005271891.1 XM_005271892.2 XM_005271894.2 XM_005271895.2 XM_005271896.1 XM_017009057.1 XM_017009058.1 XM_017009059.1 XM_017009060.1 XM_017009061.1 XM_017009062.1 XM_017009063.1 XM_017009064.1 XM_017009065.1 XM_017009066.1 XM_017009067.1 XM_017009068.1 XM_017009069.1 XM_017009070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7235962 c.236G>A p.R79K Chr.5: 123557511-123557511 on Build GRCh38
4478568 c.224C>T p.P75L Chr.5: 123557499-123557499 on Build GRCh38
1432720 c.224C>A p.P75H Chr.5: 123557499-123557499 on Build GRCh38
7128251 c.267C>G p.F89L Chr.5: 123557542-123557542 on Build GRCh38
1235998 c.279A>T p.L93F Chr.5: 123557554-123557554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753732759 Chr.5: 123557490-123557490 on Build GRCh38
rs757935911 Chr.5: 123557552-123557552 on Build GRCh38
rs571743573 Chr.5: 123557576-123557576 on Build GRCh38
rs768974149 Chr.5: 123557587-123557587 on Build GRCh38
rs761623683 Chr.5: 123557567-123557568 on Build GRCh38
rs966893598 Chr.5: 123557644-123557644 on Build GRCh38
rs113119893 Chr.5: 123557605-123557605 on Build GRCh38
rs1021245773 Chr.5: 123557636-123557636 on Build GRCh38
rs777358192 Chr.5: 123557557-123557557 on Build GRCh38
rs778923602 Chr.5: 123557539-123557539 on Build GRCh38
rs761025228 Chr.5: 123557467-123557467 on Build GRCh38
rs750693935 Chr.5: 123557461-123557461 on Build GRCh38
rs932762002 Chr.5: 123557482-123557482 on Build GRCh38
rs17854498 Chr.5: 123557546-123557546 on Build GRCh38
rs374465627 Chr.5: 123557460-123557460 on Build GRCh38
rs780817281 Chr.5: 123557563-123557563 on Build GRCh38
rs368370204 Chr.5: 123557571-123557571 on Build GRCh38
rs761985676 Chr.5: 123557447-123557447 on Build GRCh38
rs752136946 Chr.5: 123557544-123557544 on Build GRCh38
rs34834293 Chr.5: 123557469-123557470 on Build GRCh38
rs766055555 Chr.5: 123557487-123557487 on Build GRCh38
rs770660931 Chr.5: 123557572-123557572 on Build GRCh38
rs767556990 Chr.5: 123557616-123557617 on Build GRCh38
rs914021702 Chr.5: 123557611-123557611 on Build GRCh38
rs761932706 Chr.5: 123557603-123557603 on Build GRCh38
rs894018748 Chr.5: 123557524-123557524 on Build GRCh38
rs754857586 Chr.5: 123557499-123557499 on Build GRCh38

View Full Product Details