Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21416489-21416762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ12
Entrez Gene ID
3768
Gene Name
potassium voltage-gated channel subfamily J member 12
Gene Aliases
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x
UniGene
Hs.200629
Gene Chromosome Location
Chr.17: 21376387-21419870 on Build GRCh38
Transcripts
NM_021012.4 XM_005256625.4 XM_011523831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
253168 c.1222G>A p.A408T Chr.17: 21416564-21416564 on Build GRCh38
5852044 c.1181G>A p.G394E Chr.17: 21416523-21416523 on Build GRCh38
3515172 c.1253G>A p.G418D Chr.17: 21416595-21416595 on Build GRCh38
3773792 c.1191C>T p.D397D Chr.17: 21416533-21416533 on Build GRCh38
4694418 c.1264C>G p.L422V Chr.17: 21416606-21416606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782786069 Chr.17: 21416615-21416615 on Build GRCh38
rs782549834 Chr.17: 21416550-21416550 on Build GRCh38
rs2917720 Chr.17: 21416548-21416548 on Build GRCh38
rs781829352 Chr.17: 21416616-21416616 on Build GRCh38
rs781879354 Chr.17: 21416662-21416662 on Build GRCh38
rs781837062 Chr.17: 21416659-21416659 on Build GRCh38
rs782706974 Chr.17: 21416621-21416621 on Build GRCh38
rs376362557 Chr.17: 21416522-21416522 on Build GRCh38
rs782148726 Chr.17: 21416543-21416543 on Build GRCh38
rs5021700 Chr.17: 21416677-21416677 on Build GRCh38
rs781854398 Chr.17: 21416658-21416658 on Build GRCh38
rs782470361 Chr.17: 21416628-21416628 on Build GRCh38
rs5021701 Chr.17: 21416681-21416681 on Build GRCh38
rs782407544 Chr.17: 21416590-21416590 on Build GRCh38
rs77266866 Chr.17: 21416597-21416597 on Build GRCh38
rs549297035 Chr.17: 21416585-21416585 on Build GRCh38
rs782464648 Chr.17: 21416519-21416519 on Build GRCh38
rs550496794 Chr.17: 21416570-21416570 on Build GRCh38
rs200772314 Chr.17: 21416561-21416561 on Build GRCh38
rs782611815 Chr.17: 21416521-21416521 on Build GRCh38
rs782314026 Chr.17: 21416639-21416639 on Build GRCh38
rs782434519 Chr.17: 21416660-21416660 on Build GRCh38
rs782145049 Chr.17: 21416675-21416675 on Build GRCh38
rs782633689 Chr.17: 21416626-21416626 on Build GRCh38
rs782562270 Chr.17: 21416624-21416624 on Build GRCh38
rs781954971 Chr.17: 21416692-21416692 on Build GRCh38
rs781800042 Chr.17: 21416569-21416569 on Build GRCh38
rs576434712 Chr.17: 21416690-21416690 on Build GRCh38
rs781862476 Chr.17: 21416571-21416571 on Build GRCh38
rs782333305 Chr.17: 21416531-21416531 on Build GRCh38
rs781872566 Chr.17: 21416695-21416695 on Build GRCh38
rs201223000 Chr.17: 21416567-21416567 on Build GRCh38
rs201975437 Chr.17: 21416604-21416604 on Build GRCh38
rs112154887 Chr.17: 21416586-21416586 on Build GRCh38
rs781950741 Chr.17: 21416670-21416670 on Build GRCh38
rs782266470 Chr.17: 21416664-21416664 on Build GRCh38
rs782715315 Chr.17: 21416547-21416547 on Build GRCh38
rs556076946 Chr.17: 21416727-21416727 on Build GRCh38
rs782091094 Chr.17: 21416565-21416565 on Build GRCh38
rs782099091 Chr.17: 21416620-21416620 on Build GRCh38
rs781949146 Chr.17: 21416691-21416691 on Build GRCh38
rs72846670 Chr.17: 21416533-21416533 on Build GRCh38
rs782558085 Chr.17: 21416663-21416663 on Build GRCh38
rs375519519 Chr.17: 21416600-21416600 on Build GRCh38
rs782586759 Chr.17: 21416581-21416581 on Build GRCh38
rs78435971 Chr.17: 21416696-21416696 on Build GRCh38
rs782206715 Chr.17: 21416683-21416683 on Build GRCh38
rs782352669 Chr.17: 21416599-21416599 on Build GRCh38
rs782059341 Chr.17: 21416601-21416601 on Build GRCh38
rs781921645 Chr.17: 21416542-21416542 on Build GRCh38
rs34762677 Chr.17: 21416665-21416665 on Build GRCh38
rs5021699 Chr.17: 21416631-21416631 on Build GRCh38
rs782396557 Chr.17: 21416603-21416603 on Build GRCh38
rs782381983 Chr.17: 21416653-21416653 on Build GRCh38
rs782122675 Chr.17: 21416614-21416614 on Build GRCh38
rs782296754 Chr.17: 21416645-21416645 on Build GRCh38
rs76506689 Chr.17: 21416643-21416643 on Build GRCh38
rs73979902 Chr.17: 21416556-21416556 on Build GRCh38
rs781929659 Chr.17: 21416674-21416674 on Build GRCh38
rs66981137 Chr.17: 21416596-21416596 on Build GRCh38
rs782595955 Chr.17: 21416575-21416575 on Build GRCh38
rs782517673 Chr.17: 21416678-21416678 on Build GRCh38
rs537708101 Chr.17: 21416709-21416709 on Build GRCh38
rs782752752 Chr.17: 21416679-21416679 on Build GRCh38
rs782252952 Chr.17: 21416518-21416518 on Build GRCh38
rs781867776 Chr.17: 21416627-21416627 on Build GRCh38
rs558105226 Chr.17: 21416682-21416682 on Build GRCh38
rs77515828 Chr.17: 21416700-21416700 on Build GRCh38
rs529907789 Chr.17: 21416512-21416512 on Build GRCh38
rs782042622 Chr.17: 21416610-21416610 on Build GRCh38
rs5021702 Chr.17: 21416713-21416713 on Build GRCh38
rs781842331 Chr.17: 21416578-21416578 on Build GRCh38
rs782171551 Chr.17: 21416635-21416635 on Build GRCh38
rs781936770 Chr.17: 21416673-21416673 on Build GRCh38
rs782045793 Chr.17: 21416681-21416681 on Build GRCh38
rs34808513 Chr.17: 21416729-21416729 on Build GRCh38
rs782391430 Chr.17: 21416668-21416668 on Build GRCh38
rs782432903 Chr.17: 21416579-21416579 on Build GRCh38
rs782067486 Chr.17: 21416657-21416657 on Build GRCh38
rs144702327 Chr.17: 21416537-21416537 on Build GRCh38
rs781916912 Chr.17: 21416686-21416686 on Build GRCh38
rs782374936 Chr.17: 21416592-21416592 on Build GRCh38
rs564486905 Chr.17: 21416557-21416557 on Build GRCh38
rs782577366 Chr.17: 21416549-21416549 on Build GRCh38
rs782576434 Chr.17: 21416511-21416511 on Build GRCh38
rs782090321 Chr.17: 21416655-21416655 on Build GRCh38
rs782154190 Chr.17: 21416694-21416694 on Build GRCh38
rs782711389 Chr.17: 21416661-21416661 on Build GRCh38
rs781808526 Chr.17: 21416623-21416623 on Build GRCh38
rs782407309 Chr.17: 21416559-21416559 on Build GRCh38
rs782112156 Chr.17: 21416691-21416691 on Build GRCh38
rs782354132 Chr.17: 21416684-21416684 on Build GRCh38
rs782746237 Chr.17: 21416538-21416538 on Build GRCh38
rs782541675 Chr.17: 21416515-21416515 on Build GRCh38
rs782649176 Chr.17: 21416553-21416553 on Build GRCh38
rs781849043 Chr.17: 21416544-21416544 on Build GRCh38
rs782185268 Chr.17: 21416558-21416558 on Build GRCh38
rs782437581 Chr.17: 21416680-21416680 on Build GRCh38
rs782768236 Chr.17: 21416560-21416560 on Build GRCh38
rs140886041 Chr.17: 21416555-21416555 on Build GRCh38
rs568552866 Chr.17: 21416572-21416572 on Build GRCh38
rs574541333 Chr.17: 21416728-21416728 on Build GRCh38
rs782731305 Chr.17: 21416689-21416689 on Build GRCh38
rs782099004 Chr.17: 21416671-21416671 on Build GRCh38
rs782375076 Chr.17: 21416534-21416534 on Build GRCh38
rs541946650 Chr.17: 21416517-21416517 on Build GRCh38
rs782193335 Chr.17: 21416687-21416687 on Build GRCh38
rs782490035 Chr.17: 21416510-21416510 on Build GRCh38
rs782691680 Chr.17: 21416693-21416693 on Build GRCh38
rs782693112 Chr.17: 21416673-21416673 on Build GRCh38
rs782362976 Chr.17: 21416656-21416656 on Build GRCh38
rs782164148 Chr.17: 21416548-21416548 on Build GRCh38
rs782525665 Chr.17: 21416546-21416546 on Build GRCh38
rs782402105 Chr.17: 21416689-21416689 on Build GRCh38
rs782021622 Chr.17: 21416666-21416666 on Build GRCh38

View Full Product Details