Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30928456-30928711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001099784.2 NM_001282351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297029 c.786G>T p.R262S Chr.16: 30928565-30928565 on Build GRCh38
1734199 c.785_786insG p.V263fs*60 Chr.16: 30928564-30928565 on Build GRCh38
6279824 c.745C>T p.R249* Chr.16: 30928524-30928524 on Build GRCh38
4919294 c.717G>A p.K239K Chr.16: 30928496-30928496 on Build GRCh38
703238 c.784A>T p.R262W Chr.16: 30928563-30928563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771879119 Chr.16: 30928665-30928665 on Build GRCh38
rs376169841 Chr.16: 30928511-30928511 on Build GRCh38
rs771051806 Chr.16: 30928576-30928576 on Build GRCh38
rs774376414 Chr.16: 30928649-30928649 on Build GRCh38
rs373602730 Chr.16: 30928542-30928542 on Build GRCh38
rs751468850 Chr.16: 30928525-30928525 on Build GRCh38
rs760293502 Chr.16: 30928615-30928615 on Build GRCh38
rs765229438 Chr.16: 30928512-30928512 on Build GRCh38
rs773309908 Chr.16: 30928513-30928513 on Build GRCh38
rs561775075 Chr.16: 30928537-30928537 on Build GRCh38
rs763558944 Chr.16: 30928621-30928621 on Build GRCh38
rs748132003 Chr.16: 30928560-30928560 on Build GRCh38
rs370574317 Chr.16: 30928611-30928611 on Build GRCh38
rs773960782 Chr.16: 30928577-30928577 on Build GRCh38
rs767211251 Chr.16: 30928531-30928531 on Build GRCh38
rs541182178 Chr.16: 30928547-30928547 on Build GRCh38
rs755679850 Chr.16: 30928535-30928535 on Build GRCh38
rs763987814 Chr.16: 30928548-30928552 on Build GRCh38
rs369163910 Chr.16: 30928524-30928524 on Build GRCh38
rs747165398 Chr.16: 30928496-30928496 on Build GRCh38
rs773465661 Chr.16: 30928570-30928570 on Build GRCh38
rs377586993 Chr.16: 30928597-30928597 on Build GRCh38
rs775878214 Chr.16: 30928491-30928491 on Build GRCh38
rs768696380 Chr.16: 30928545-30928545 on Build GRCh38
rs781462362 Chr.16: 30928641-30928641 on Build GRCh38
rs762980615 Chr.16: 30928571-30928571 on Build GRCh38
rs770923391 Chr.16: 30928648-30928648 on Build GRCh38
rs769976000 Chr.16: 30928561-30928561 on Build GRCh38
rs748372068 Chr.16: 30928642-30928642 on Build GRCh38
rs200337123 Chr.16: 30928480-30928480 on Build GRCh38
rs770753963 Chr.16: 30928538-30928538 on Build GRCh38
rs371190776 Chr.16: 30928520-30928520 on Build GRCh38
rs746191715 Chr.16: 30928479-30928479 on Build GRCh38
rs770209716 Chr.16: 30928644-30928644 on Build GRCh38
rs759202048 Chr.16: 30928652-30928652 on Build GRCh38
rs895449292 Chr.16: 30928667-30928667 on Build GRCh38
rs752080541 Chr.16: 30928532-30928532 on Build GRCh38
rs373190519 Chr.16: 30928503-30928503 on Build GRCh38
rs376320900 Chr.16: 30928541-30928541 on Build GRCh38
rs752416227 Chr.16: 30928614-30928614 on Build GRCh38
rs749394811 Chr.16: 30928647-30928647 on Build GRCh38
rs776243326 Chr.16: 30928635-30928635 on Build GRCh38
rs201671986 Chr.16: 30928676-30928676 on Build GRCh38
rs375812010 Chr.16: 30928634-30928634 on Build GRCh38
rs373879470 Chr.16: 30928523-30928523 on Build GRCh38
rs753410617 Chr.16: 30928629-30928629 on Build GRCh38
rs759377390 Chr.16: 30928589-30928589 on Build GRCh38
rs747462560 Chr.16: 30928543-30928543 on Build GRCh38
rs767036076 Chr.16: 30928591-30928591 on Build GRCh38
rs768986133 Chr.16: 30928502-30928502 on Build GRCh38
rs202132610 Chr.16: 30928546-30928546 on Build GRCh38
rs902924531 Chr.16: 30928623-30928623 on Build GRCh38
rs777988743 Chr.16: 30928645-30928645 on Build GRCh38
rs755396814 Chr.16: 30928637-30928637 on Build GRCh38
rs760312482 Chr.16: 30928677-30928677 on Build GRCh38
rs368260277 Chr.16: 30928636-30928636 on Build GRCh38

View Full Product Details