Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67178480-67178715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895L
Entrez Gene ID
653319
Gene Name
KIAA0895 like
Gene Aliases
-
UniGene
Hs.651201
Gene Chromosome Location
Chr.16: 67175602-67183980 on Build GRCh38
Transcripts
NM_001040715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5033124 c.298C>G p.P100A Chr.16: 67178589-67178589 on Build GRCh38
7251929 c.282G>A p.S94S Chr.16: 67178605-67178605 on Build GRCh38
6283918 c.228G>A p.G76G Chr.16: 67178659-67178659 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs998932382 Chr.16: 67178653-67178653 on Build GRCh38
rs948739740 Chr.16: 67178610-67178610 on Build GRCh38
rs760047481 Chr.16: 67178664-67178664 on Build GRCh38
rs779626606 Chr.16: 67178505-67178505 on Build GRCh38
rs1032277096 Chr.16: 67178676-67178676 on Build GRCh38
rs922387941 Chr.16: 67178517-67178517 on Build GRCh38
rs373857221 Chr.16: 67178647-67178647 on Build GRCh38
rs933740872 Chr.16: 67178520-67178520 on Build GRCh38
rs553620572 Chr.16: 67178617-67178617 on Build GRCh38
rs540684396 Chr.16: 67178537-67178537 on Build GRCh38
rs941287393 Chr.16: 67178598-67178598 on Build GRCh38
rs1017775917 Chr.16: 67178690-67178690 on Build GRCh38
rs114286521 Chr.16: 67178604-67178604 on Build GRCh38
rs1011794330 Chr.16: 67178684-67178684 on Build GRCh38
rs746679675 Chr.16: 67178593-67178593 on Build GRCh38
rs772560672 Chr.16: 67178575-67178575 on Build GRCh38
rs776334913 Chr.16: 67178622-67178622 on Build GRCh38
rs964921653 Chr.16: 67178696-67178696 on Build GRCh38
rs894482007 Chr.16: 67178607-67178607 on Build GRCh38
rs983043827 Chr.16: 67178547-67178547 on Build GRCh38
rs528425637 Chr.16: 67178613-67178613 on Build GRCh38

View Full Product Details