Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68952727-68952996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKN2
Entrez Gene ID
200504
Gene Name
gastrokine 2
Gene Aliases
BRICD1B, GDDR, PRO813, TFIZ1, VLTI465
UniGene
Hs.16757
Gene Chromosome Location
Chr.2: 68945232-68952970 on Build GRCh38
Transcripts
NM_182536.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671333 c.5A>C p.K2T Chr.2: 68952857-68952857 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs985357992 Chr.2: 68952936-68952936 on Build GRCh38
rs910169622 Chr.2: 68952794-68952794 on Build GRCh38
rs758067741 Chr.2: 68952873-68952873 on Build GRCh38
rs747706227 Chr.2: 68952908-68952908 on Build GRCh38
rs965361811 Chr.2: 68952948-68952948 on Build GRCh38
rs1018298290 Chr.2: 68952753-68952756 on Build GRCh38
rs963415919 Chr.2: 68952883-68952883 on Build GRCh38
rs746997747 Chr.2: 68952782-68952782 on Build GRCh38
rs1019939537 Chr.2: 68952939-68952939 on Build GRCh38
rs6720316 Chr.2: 68952842-68952842 on Build GRCh38
rs74619915 Chr.2: 68952808-68952808 on Build GRCh38
rs745523689 Chr.2: 68952840-68952840 on Build GRCh38
rs192229708 Chr.2: 68952815-68952815 on Build GRCh38
rs772718036 Chr.2: 68952832-68952832 on Build GRCh38
rs989031780 Chr.2: 68952961-68952961 on Build GRCh38
rs570989394 Chr.2: 68952778-68952778 on Build GRCh38
rs200100367 Chr.2: 68952899-68952899 on Build GRCh38
rs540901512 Chr.2: 68952843-68952843 on Build GRCh38
rs779699264 Chr.2: 68952881-68952881 on Build GRCh38
rs752781443 Chr.2: 68952852-68952852 on Build GRCh38
rs760085849 Chr.2: 68952927-68952927 on Build GRCh38
rs777064567 Chr.2: 68952864-68952864 on Build GRCh38
rs535123087 Chr.2: 68952821-68952821 on Build GRCh38
rs763288253 Chr.2: 68952853-68952853 on Build GRCh38
rs532910142 Chr.2: 68952820-68952820 on Build GRCh38
rs965877262 Chr.2: 68952760-68952760 on Build GRCh38
rs200894742 Chr.2: 68952858-68952858 on Build GRCh38
rs780076241 Chr.2: 68952800-68952800 on Build GRCh38
rs140417428 Chr.2: 68952779-68952779 on Build GRCh38
rs370746418 Chr.2: 68952845-68952845 on Build GRCh38
rs756003255 Chr.2: 68952876-68952876 on Build GRCh38
rs6720209 Chr.2: 68952769-68952769 on Build GRCh38
rs1028926912 Chr.2: 68952923-68952923 on Build GRCh38
rs765414650 Chr.2: 68952847-68952847 on Build GRCh38
rs778419436 Chr.2: 68952900-68952900 on Build GRCh38
rs753825871 Chr.2: 68952897-68952897 on Build GRCh38
rs749176816 Chr.2: 68952807-68952807 on Build GRCh38
rs75502905 Chr.2: 68952837-68952837 on Build GRCh38

View Full Product Details