Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2029566-2029825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRD
Entrez Gene ID
2563
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Gene Aliases
EIG10, EJM7, GEFSP5
UniGene
Hs.113882
Gene Chromosome Location
Chr.1: 2019329-2030753 on Build GRCh38
Transcripts
NM_000815.4 XM_011541194.2 XM_017000936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668638 c.910G>A p.A304T Chr.1: 2029613-2029613 on Build GRCh38
463652 c.968C>T p.A323V Chr.1: 2029671-2029671 on Build GRCh38
901486 c.1017G>T p.K339N Chr.1: 2029720-2029720 on Build GRCh38
4026443 c.1049C>T p.P350L Chr.1: 2029752-2029752 on Build GRCh38
2124564 c.960C>T p.F320F Chr.1: 2029663-2029663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576082385 Chr.1: 2029765-2029765 on Build GRCh38
rs541562450 Chr.1: 2029778-2029778 on Build GRCh38
rs200124298 Chr.1: 2029636-2029636 on Build GRCh38
rs896016478 Chr.1: 2029673-2029673 on Build GRCh38
rs1036762635 Chr.1: 2029681-2029681 on Build GRCh38
rs759215834 Chr.1: 2029752-2029752 on Build GRCh38
rs775080616 Chr.1: 2029803-2029803 on Build GRCh38
rs781426402 Chr.1: 2029611-2029611 on Build GRCh38
rs376640236 Chr.1: 2029666-2029666 on Build GRCh38
rs770693073 Chr.1: 2029796-2029796 on Build GRCh38
rs749316684 Chr.1: 2029730-2029730 on Build GRCh38
rs767909003 Chr.1: 2029773-2029773 on Build GRCh38
rs146381127 Chr.1: 2029672-2029672 on Build GRCh38
rs201101812 Chr.1: 2029663-2029663 on Build GRCh38
rs762485697 Chr.1: 2029806-2029806 on Build GRCh38
rs769505409 Chr.1: 2029802-2029802 on Build GRCh38
rs759009305 Chr.1: 2029687-2029687 on Build GRCh38
rs555522768 Chr.1: 2029786-2029786 on Build GRCh38
rs368765147 Chr.1: 2029675-2029675 on Build GRCh38
rs776198921 Chr.1: 2029747-2029747 on Build GRCh38
rs780175857 Chr.1: 2029729-2029729 on Build GRCh38
rs866697106 Chr.1: 2029719-2029719 on Build GRCh38
rs764783225 Chr.1: 2029704-2029704 on Build GRCh38
rs762397628 Chr.1: 2029756-2029756 on Build GRCh38
rs139676179 Chr.1: 2029645-2029645 on Build GRCh38
rs41315330 Chr.1: 2029705-2029705 on Build GRCh38
rs141082183 Chr.1: 2029742-2029742 on Build GRCh38
rs777901210 Chr.1: 2029788-2029788 on Build GRCh38
rs757601943 Chr.1: 2029717-2029717 on Build GRCh38
rs773238325 Chr.1: 2029735-2029735 on Build GRCh38
rs201728821 Chr.1: 2029753-2029753 on Build GRCh38
rs757483435 Chr.1: 2029588-2029588 on Build GRCh38
rs1046709660 Chr.1: 2029726-2029726 on Build GRCh38
rs142099913 Chr.1: 2029732-2029732 on Build GRCh38
rs772061844 Chr.1: 2029653-2029653 on Build GRCh38
rs758908317 Chr.1: 2029706-2029706 on Build GRCh38
rs200391871 Chr.1: 2029595-2029595 on Build GRCh38
rs780367048 Chr.1: 2029779-2029779 on Build GRCh38
rs937477150 Chr.1: 2029669-2029669 on Build GRCh38
rs904696266 Chr.1: 2029754-2029755 on Build GRCh38
rs750676768 Chr.1: 2029775-2029775 on Build GRCh38
rs774201718 Chr.1: 2029637-2029637 on Build GRCh38
rs966994022 Chr.1: 2029805-2029805 on Build GRCh38
rs948197538 Chr.1: 2029649-2029649 on Build GRCh38
rs757611508 Chr.1: 2029771-2029771 on Build GRCh38
rs752030520 Chr.1: 2029770-2029770 on Build GRCh38
rs28507434 Chr.1: 2029800-2029800 on Build GRCh38
rs746924719 Chr.1: 2029789-2029789 on Build GRCh38
rs746076615 Chr.1: 2029718-2029718 on Build GRCh38
rs370201162 Chr.1: 2029708-2029708 on Build GRCh38
rs746832451 Chr.1: 2029736-2029736 on Build GRCh38
rs866215877 Chr.1: 2029613-2029613 on Build GRCh38
rs753995198 Chr.1: 2029783-2029783 on Build GRCh38
rs756202555 Chr.1: 2029727-2029727 on Build GRCh38
rs745961044 Chr.1: 2029631-2029631 on Build GRCh38
rs781038575 Chr.1: 2029799-2029799 on Build GRCh38

View Full Product Details