Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6015290-6015563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLS1
Entrez Gene ID
54675
Gene Name
cardiolipin synthase 1
Gene Aliases
C20orf155, CLS, CLS1, GCD10, dJ967N21.6
UniGene
Hs.743956
Gene Chromosome Location
Chr.20: 6005938-6040053 on Build GRCh38
Transcripts
NM_001127458.1 NM_001323561.1 NM_001323562.1 NM_001323563.1 NM_001323564.1 NM_019095.5 NR_136617.1 XM_005260738.4 XM_011529263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1028738 c.555C>A p.T185T Chr.20: 6015471-6015471 on Build GRCh38
5707501 c.466A>G p.N156D Chr.20: 6015382-6015382 on Build GRCh38
3405240 c.562G>T p.D188Y Chr.20: 6015478-6015478 on Build GRCh38
7232268 c.181C>G p.Q61E Chr.20: 6015394-6015394 on Build GRCh38
7285687 c.574+9G>A p.? Chr.20: 6015499-6015499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769793707 Chr.20: 6015445-6015445 on Build GRCh38
rs777538525 Chr.20: 6015504-6015504 on Build GRCh38
rs376164397 Chr.20: 6015501-6015501 on Build GRCh38
rs776696810 Chr.20: 6015404-6015404 on Build GRCh38
rs560844185 Chr.20: 6015508-6015508 on Build GRCh38
rs747602046 Chr.20: 6015351-6015351 on Build GRCh38
rs765446271 Chr.20: 6015339-6015340 on Build GRCh38
rs199865968 Chr.20: 6015338-6015339 on Build GRCh38
rs754166011 Chr.20: 6015490-6015490 on Build GRCh38
rs759395960 Chr.20: 6015431-6015431 on Build GRCh38
rs1027121031 Chr.20: 6015454-6015454 on Build GRCh38
rs367625466 Chr.20: 6015389-6015389 on Build GRCh38
rs752237007 Chr.20: 6015500-6015500 on Build GRCh38
rs773286368 Chr.20: 6015456-6015456 on Build GRCh38
rs745605369 Chr.20: 6015533-6015533 on Build GRCh38
rs890793874 Chr.20: 6015344-6015344 on Build GRCh38
rs757131180 Chr.20: 6015511-6015512 on Build GRCh38
rs755489546 Chr.20: 6015498-6015498 on Build GRCh38
rs780953282 Chr.20: 6015474-6015474 on Build GRCh38
rs761941118 Chr.20: 6015446-6015446 on Build GRCh38
rs746637754 Chr.20: 6015507-6015507 on Build GRCh38
rs745571120 Chr.20: 6015499-6015499 on Build GRCh38
rs759697008 Chr.20: 6015339-6015339 on Build GRCh38
rs369155508 Chr.20: 6015380-6015380 on Build GRCh38
rs771735217 Chr.20: 6015353-6015353 on Build GRCh38
rs766652713 Chr.20: 6015473-6015473 on Build GRCh38
rs373154036 Chr.20: 6015470-6015470 on Build GRCh38
rs754289374 Chr.20: 6015340-6015340 on Build GRCh38
rs760651040 Chr.20: 6015327-6015330 on Build GRCh38
rs756406802 Chr.20: 6015336-6015336 on Build GRCh38
rs181766120 Chr.20: 6015339-6015339 on Build GRCh38
rs757078288 Chr.20: 6015347-6015347 on Build GRCh38
rs777252451 Chr.20: 6015361-6015361 on Build GRCh38
rs542675915 Chr.20: 6015346-6015346 on Build GRCh38
rs767895881 Chr.20: 6015452-6015452 on Build GRCh38
rs766440895 Chr.20: 6015329-6015330 on Build GRCh38
rs750827604 Chr.20: 6015332-6015332 on Build GRCh38
rs371623502 Chr.20: 6015525-6015525 on Build GRCh38
rs751348675 Chr.20: 6015470-6015486 on Build GRCh38

View Full Product Details