Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154611993-154612220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGG
Entrez Gene ID
2266
Gene Name
fibrinogen gamma chain
Gene Aliases
-
UniGene
Hs.727584
Gene Chromosome Location
Chr.4: 154604134-154612808 on Build GRCh38
Transcripts
NM_000509.5 NM_021870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5934875 c.227T>G p.V76G Chr.4: 154612098-154612098 on Build GRCh38
3129325 c.187G>C p.D63H Chr.4: 154612138-154612138 on Build GRCh38
3775644 c.296C>T p.S99L Chr.4: 154612029-154612029 on Build GRCh38
4684629 c.203C>A p.S68Y Chr.4: 154612122-154612122 on Build GRCh38
1739042 c.124G>A p.G42S Chr.4: 154612201-154612201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781702629 Chr.4: 154612148-154612148 on Build GRCh38
rs770096313 Chr.4: 154612045-154612045 on Build GRCh38
rs769139316 Chr.4: 154612023-154612023 on Build GRCh38
rs766605366 Chr.4: 154612050-154612050 on Build GRCh38
rs142286849 Chr.4: 154612051-154612051 on Build GRCh38
rs918243438 Chr.4: 154612100-154612100 on Build GRCh38
rs771243311 Chr.4: 154612179-154612179 on Build GRCh38
rs748525533 Chr.4: 154612044-154612044 on Build GRCh38
rs748671641 Chr.4: 154612155-154612155 on Build GRCh38
rs11551850 Chr.4: 154612064-154612064 on Build GRCh38
rs11551835 Chr.4: 154612095-154612095 on Build GRCh38
rs754497653 Chr.4: 154612138-154612138 on Build GRCh38
rs138511699 Chr.4: 154612185-154612185 on Build GRCh38
rs142224295 Chr.4: 154612112-154612112 on Build GRCh38
rs757041089 Chr.4: 154612104-154612104 on Build GRCh38
rs756652149 Chr.4: 154612047-154612047 on Build GRCh38
rs778057919 Chr.4: 154612172-154612172 on Build GRCh38
rs369888820 Chr.4: 154612049-154612049 on Build GRCh38
rs376769607 Chr.4: 154612129-154612129 on Build GRCh38
rs779592161 Chr.4: 154612125-154612125 on Build GRCh38
rs967471536 Chr.4: 154612145-154612145 on Build GRCh38
rs777027712 Chr.4: 154612038-154612038 on Build GRCh38
rs764040983 Chr.4: 154612077-154612077 on Build GRCh38
rs563975687 Chr.4: 154612130-154612130 on Build GRCh38
rs750086605 Chr.4: 154612123-154612123 on Build GRCh38
rs770477991 Chr.4: 154612144-154612144 on Build GRCh38
rs11551840 Chr.4: 154612165-154612165 on Build GRCh38
rs753766294 Chr.4: 154612102-154612102 on Build GRCh38
rs770184920 Chr.4: 154612163-154612163 on Build GRCh38
rs145914446 Chr.4: 154612136-154612136 on Build GRCh38
rs11551836 Chr.4: 154612169-154612169 on Build GRCh38
rs11551849 Chr.4: 154612059-154612059 on Build GRCh38
rs778213856 Chr.4: 154612072-154612072 on Build GRCh38

View Full Product Details