Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70656922-70657189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
UniGene
Hs.461214
Gene Chromosome Location
Chr.16: 70579895-70660682 on Build GRCh38
Transcripts
NM_001172771.1 NM_001172772.1 NM_152456.2 XM_011522902.2 XM_011522903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685337 c.386T>C p.V129A Chr.16: 70657105-70657105 on Build GRCh38
3273311 c.372G>A p.T124T Chr.16: 70657091-70657091 on Build GRCh38
4062386 c.356T>C p.L119P Chr.16: 70657075-70657075 on Build GRCh38
6685332 c.321C>T p.D107D Chr.16: 70657040-70657040 on Build GRCh38
6685333 c.330C>T p.L110L Chr.16: 70657049-70657049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8046424 Chr.16: 70657086-70657086 on Build GRCh38
rs202131686 Chr.16: 70656977-70656977 on Build GRCh38
rs754023755 Chr.16: 70657162-70657162 on Build GRCh38
rs114319372 Chr.16: 70657147-70657147 on Build GRCh38
rs767053852 Chr.16: 70656957-70656957 on Build GRCh38
rs770414893 Chr.16: 70657114-70657114 on Build GRCh38
rs755199626 Chr.16: 70657085-70657085 on Build GRCh38
rs763961200 Chr.16: 70657137-70657137 on Build GRCh38
rs368923655 Chr.16: 70657120-70657120 on Build GRCh38
rs760301983 Chr.16: 70656978-70656978 on Build GRCh38
rs769134037 Chr.16: 70657022-70657022 on Build GRCh38
rs572668662 Chr.16: 70656990-70656990 on Build GRCh38
rs374730889 Chr.16: 70657028-70657028 on Build GRCh38
rs374324186 Chr.16: 70657031-70657031 on Build GRCh38
rs374665339 Chr.16: 70657060-70657060 on Build GRCh38
rs952622576 Chr.16: 70657019-70657019 on Build GRCh38
rs763895154 Chr.16: 70657050-70657050 on Build GRCh38
rs760707763 Chr.16: 70657164-70657177 on Build GRCh38
rs78909989 Chr.16: 70656955-70656955 on Build GRCh38
rs781308018 Chr.16: 70656991-70656991 on Build GRCh38
rs77769000 Chr.16: 70657013-70657013 on Build GRCh38
rs780354617 Chr.16: 70657007-70657007 on Build GRCh38
rs1024630832 Chr.16: 70656971-70656971 on Build GRCh38
rs771370857 Chr.16: 70657041-70657042 on Build GRCh38
rs778076818 Chr.16: 70657095-70657095 on Build GRCh38
rs771623956 Chr.16: 70657150-70657151 on Build GRCh38
rs541388831 Chr.16: 70657160-70657160 on Build GRCh38
rs774848853 Chr.16: 70657152-70657153 on Build GRCh38
rs780334521 Chr.16: 70657065-70657065 on Build GRCh38
rs573956551 Chr.16: 70657104-70657104 on Build GRCh38
rs751544519 Chr.16: 70657051-70657051 on Build GRCh38
rs756291278 Chr.16: 70657006-70657006 on Build GRCh38
rs142587592 Chr.16: 70657049-70657049 on Build GRCh38
rs765338615 Chr.16: 70657166-70657166 on Build GRCh38
rs759042156 Chr.16: 70657151-70657152 on Build GRCh38
rs368553371 Chr.16: 70657040-70657040 on Build GRCh38
rs766539623 Chr.16: 70657154-70657154 on Build GRCh38
rs755110920 Chr.16: 70657165-70657165 on Build GRCh38
rs200597979 Chr.16: 70657057-70657057 on Build GRCh38
rs371553529 Chr.16: 70657121-70657121 on Build GRCh38
rs370436386 Chr.16: 70657024-70657024 on Build GRCh38
rs772518810 Chr.16: 70657091-70657091 on Build GRCh38
rs1033503960 Chr.16: 70657014-70657014 on Build GRCh38
rs915734607 Chr.16: 70657054-70657054 on Build GRCh38
rs949962962 Chr.16: 70657071-70657071 on Build GRCh38
rs762585609 Chr.16: 70657131-70657131 on Build GRCh38
rs368143418 Chr.16: 70657030-70657030 on Build GRCh38
rs976274551 Chr.16: 70657021-70657021 on Build GRCh38
rs761912414 Chr.16: 70657152-70657152 on Build GRCh38
rs372238545 Chr.16: 70657044-70657044 on Build GRCh38
rs774176294 Chr.16: 70657134-70657135 on Build GRCh38
rs75180225 Chr.16: 70657168-70657168 on Build GRCh38
rs776158317 Chr.16: 70657115-70657115 on Build GRCh38
rs779215796 Chr.16: 70657023-70657023 on Build GRCh38
rs377411431 Chr.16: 70656982-70656982 on Build GRCh38
rs747528604 Chr.16: 70657113-70657113 on Build GRCh38
rs376056831 Chr.16: 70657127-70657127 on Build GRCh38
rs201477178 Chr.16: 70656956-70656956 on Build GRCh38
rs770433129 Chr.16: 70657034-70657034 on Build GRCh38
rs769144263 Chr.16: 70656943-70656943 on Build GRCh38
rs779691091 Chr.16: 70656981-70656981 on Build GRCh38
rs769750203 Chr.16: 70657090-70657090 on Build GRCh38
rs754112108 Chr.16: 70657073-70657073 on Build GRCh38
rs1049053187 Chr.16: 70657094-70657094 on Build GRCh38
rs759251394 Chr.16: 70657041-70657041 on Build GRCh38

View Full Product Details