Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78013067-78013328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB4
Entrez Gene ID
11080
Gene Name
DnaJ heat shock protein family (Hsp40) member B4
Gene Aliases
DNAJW, DjB4, HLJ1
UniGene
Hs.13852
Gene Chromosome Location
Chr.1: 77979158-78017964 on Build GRCh38
Transcripts
NM_001317099.1 NM_001317100.1 NM_001317101.1 NM_001317102.1 NM_001317103.1 NM_007034.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4488502 c.333C>T p.F111F Chr.1: 78013172-78013172 on Build GRCh38
4484515 c.281C>T p.P94L Chr.1: 78013120-78013120 on Build GRCh38
3930859 c.462C>T p.L154L Chr.1: 78013301-78013301 on Build GRCh38
426715 c.375G>A p.M125I Chr.1: 78013214-78013214 on Build GRCh38
4010011 c.343C>T p.R115* Chr.1: 78013182-78013182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572512049 Chr.1: 78013287-78013287 on Build GRCh38
rs755180428 Chr.1: 78013115-78013115 on Build GRCh38
rs377300703 Chr.1: 78013094-78013094 on Build GRCh38
rs762675814 Chr.1: 78013177-78013177 on Build GRCh38
rs971788637 Chr.1: 78013236-78013236 on Build GRCh38
rs779175704 Chr.1: 78013259-78013259 on Build GRCh38
rs138757165 Chr.1: 78013219-78013219 on Build GRCh38
rs112191937 Chr.1: 78013196-78013196 on Build GRCh38
rs752918690 Chr.1: 78013281-78013281 on Build GRCh38
rs756468720 Chr.1: 78013098-78013098 on Build GRCh38
rs769857390 Chr.1: 78013156-78013156 on Build GRCh38
rs780333619 Chr.1: 78013100-78013100 on Build GRCh38
rs768311310 Chr.1: 78013183-78013183 on Build GRCh38
rs761375819 Chr.1: 78013206-78013206 on Build GRCh38
rs188409967 Chr.1: 78013297-78013297 on Build GRCh38
rs773911680 Chr.1: 78013192-78013192 on Build GRCh38
rs745865013 Chr.1: 78013145-78013145 on Build GRCh38
rs748229141 Chr.1: 78013122-78013122 on Build GRCh38
rs540017433 Chr.1: 78013296-78013296 on Build GRCh38
rs769068282 Chr.1: 78013146-78013146 on Build GRCh38
rs908423114 Chr.1: 78013194-78013194 on Build GRCh38
rs772091593 Chr.1: 78013130-78013130 on Build GRCh38
rs971448096 Chr.1: 78013189-78013189 on Build GRCh38
rs749944596 Chr.1: 78013225-78013225 on Build GRCh38
rs267598730 Chr.1: 78013204-78013204 on Build GRCh38
rs146237619 Chr.1: 78013231-78013231 on Build GRCh38
rs755340728 Chr.1: 78013254-78013254 on Build GRCh38
rs779035311 Chr.1: 78013116-78013116 on Build GRCh38
rs775364732 Chr.1: 78013172-78013172 on Build GRCh38
rs754161604 Chr.1: 78013251-78013251 on Build GRCh38
rs766895359 Chr.1: 78013248-78013248 on Build GRCh38
rs201202841 Chr.1: 78013137-78013137 on Build GRCh38
rs749667330 Chr.1: 78013112-78013112 on Build GRCh38

View Full Product Details