Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219418917-219419182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DES
Entrez Gene ID
1674
Gene Name
desmin
Gene Aliases
CSM1, CSM2, LGMD2R
UniGene
Hs.594952
Gene Chromosome Location
Chr.2: 219418377-219426739 on Build GRCh38
Transcripts
NM_001927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4677118 c.465C>T p.Y155Y Chr.2: 219418927-219418927 on Build GRCh38
3391493 c.523C>T p.R175C Chr.2: 219418985-219418985 on Build GRCh38
6026513 c.578+10C>T p.? Chr.2: 219419050-219419050 on Build GRCh38
1203407 c.479G>A p.R160Q Chr.2: 219418941-219418941 on Build GRCh38
6317785 c.459G>T p.E153D Chr.2: 219418921-219418921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014762661 Chr.2: 219418978-219418978 on Build GRCh38
rs754033658 Chr.2: 219419058-219419058 on Build GRCh38
rs907677 Chr.2: 219419104-219419104 on Build GRCh38
rs762561 Chr.2: 219419122-219419122 on Build GRCh38
rs889191100 Chr.2: 219419037-219419037 on Build GRCh38
rs969392028 Chr.2: 219419050-219419050 on Build GRCh38
rs878854473 Chr.2: 219419018-219419018 on Build GRCh38
rs961874998 Chr.2: 219418980-219418980 on Build GRCh38
rs534000139 Chr.2: 219419135-219419135 on Build GRCh38
rs757644636 Chr.2: 219419002-219419002 on Build GRCh38
rs769277716 Chr.2: 219419075-219419075 on Build GRCh38
rs747420535 Chr.2: 219419001-219419001 on Build GRCh38
rs746437365 Chr.2: 219419071-219419071 on Build GRCh38
rs878854472 Chr.2: 219418986-219418986 on Build GRCh38
rs769096434 Chr.2: 219418978-219418979 on Build GRCh38
rs1025323214 Chr.2: 219419028-219419028 on Build GRCh38
rs752944882 Chr.2: 219418979-219418979 on Build GRCh38
rs2854888 Chr.2: 219419158-219419158 on Build GRCh38
rs111548596 Chr.2: 219419051-219419051 on Build GRCh38
rs1050510822 Chr.2: 219419014-219419014 on Build GRCh38
rs958109286 Chr.2: 219419138-219419138 on Build GRCh38
rs886044489 Chr.2: 219419029-219419029 on Build GRCh38
rs538229035 Chr.2: 219418982-219418982 on Build GRCh38
rs139948855 Chr.2: 219419145-219419145 on Build GRCh38
rs1038211323 Chr.2: 219419074-219419074 on Build GRCh38
rs758686466 Chr.2: 219418981-219418981 on Build GRCh38

View Full Product Details