Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50713112-50713385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU7
Entrez Gene ID
220107
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
-
UniGene
Hs.673860
Gene Chromosome Location
Chr.13: 50711026-50843939 on Build GRCh38
Transcripts
NM_198989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203691 c.473C>T p.A158V Chr.13: 50713227-50713227 on Build GRCh38
6895045 c.482G>A p.*161* Chr.13: 50713218-50713218 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558407787 Chr.13: 50713211-50713211 on Build GRCh38
rs926089464 Chr.13: 50713258-50713258 on Build GRCh38
rs570509528 Chr.13: 50713251-50713251 on Build GRCh38
rs1046080373 Chr.13: 50713332-50713332 on Build GRCh38
rs745945052 Chr.13: 50713225-50713225 on Build GRCh38
rs369301790 Chr.13: 50713228-50713228 on Build GRCh38
rs761536983 Chr.13: 50713278-50713278 on Build GRCh38
rs750379089 Chr.13: 50713168-50713168 on Build GRCh38
rs778536502 Chr.13: 50713198-50713198 on Build GRCh38
rs753628246 Chr.13: 50713176-50713176 on Build GRCh38
rs41284836 Chr.13: 50713272-50713272 on Build GRCh38
rs778007550 Chr.13: 50713343-50713343 on Build GRCh38
rs754827956 Chr.13: 50713191-50713191 on Build GRCh38
rs746333844 Chr.13: 50713282-50713283 on Build GRCh38
rs752860836 Chr.13: 50713307-50713307 on Build GRCh38
rs756183917 Chr.13: 50713317-50713317 on Build GRCh38
rs560928686 Chr.13: 50713195-50713195 on Build GRCh38
rs944992432 Chr.13: 50713327-50713327 on Build GRCh38
rs767679094 Chr.13: 50713302-50713302 on Build GRCh38
rs750400267 Chr.13: 50713188-50713188 on Build GRCh38
rs376901047 Chr.13: 50713215-50713215 on Build GRCh38
rs781308560 Chr.13: 50713219-50713219 on Build GRCh38
rs770324729 Chr.13: 50713235-50713235 on Build GRCh38
rs781413764 Chr.13: 50713150-50713150 on Build GRCh38
rs747260789 Chr.13: 50713207-50713207 on Build GRCh38
rs370851244 Chr.13: 50713281-50713281 on Build GRCh38
rs372691233 Chr.13: 50713146-50713146 on Build GRCh38
rs937758007 Chr.13: 50713350-50713350 on Build GRCh38

View Full Product Details