Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26340110-26340383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR2
Entrez Gene ID
3709
Gene Name
inositol 1,4,5-trisphosphate receptor type 2
Gene Aliases
ANHD, CFAP48, INSP3R2, IP3R2
UniGene
Hs.512235
Gene Chromosome Location
Chr.12: 26335337-26833198 on Build GRCh38
Transcripts
NM_002223.3 XM_017019266.1 XM_017019267.1 XM_017019268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6688774 c.8004G>A p.A2668A Chr.12: 26340182-26340182 on Build GRCh38
2202846 c.7888C>T p.R2630* Chr.12: 26340298-26340298 on Build GRCh38
282027 c.7939C>T p.R2647W Chr.12: 26340247-26340247 on Build GRCh38
7372754 c.8002G>A p.A2668T Chr.12: 26340184-26340184 on Build GRCh38
275605 c.7891G>A p.A2631T Chr.12: 26340295-26340295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961083456 Chr.12: 26340136-26340136 on Build GRCh38
rs777797914 Chr.12: 26340182-26340182 on Build GRCh38
rs979025867 Chr.12: 26340344-26340344 on Build GRCh38
rs780450499 Chr.12: 26340152-26340152 on Build GRCh38
rs1034018122 Chr.12: 26340223-26340223 on Build GRCh38
rs762953295 Chr.12: 26340244-26340244 on Build GRCh38
rs959703425 Chr.12: 26340241-26340241 on Build GRCh38
rs765771678 Chr.12: 26340160-26340160 on Build GRCh38
rs540971850 Chr.12: 26340268-26340268 on Build GRCh38
rs775988616 Chr.12: 26340243-26340243 on Build GRCh38
rs757835868 Chr.12: 26340151-26340151 on Build GRCh38
rs150397176 Chr.12: 26340350-26340350 on Build GRCh38
rs771814238 Chr.12: 26340134-26340134 on Build GRCh38
rs779886485 Chr.12: 26340193-26340193 on Build GRCh38
rs752731482 Chr.12: 26340162-26340162 on Build GRCh38
rs749145663 Chr.12: 26340194-26340194 on Build GRCh38
rs778938539 Chr.12: 26340287-26340287 on Build GRCh38
rs61757116 Chr.12: 26340184-26340184 on Build GRCh38
rs542668536 Chr.12: 26340356-26340358 on Build GRCh38
rs967130602 Chr.12: 26340323-26340323 on Build GRCh38
rs754209526 Chr.12: 26340159-26340159 on Build GRCh38
rs748105897 Chr.12: 26340303-26340303 on Build GRCh38
rs760358006 Chr.12: 26340221-26340221 on Build GRCh38
rs771629804 Chr.12: 26340217-26340217 on Build GRCh38
rs757479030 Chr.12: 26340269-26340269 on Build GRCh38
rs914724710 Chr.12: 26340331-26340331 on Build GRCh38
rs754845715 Chr.12: 26340284-26340284 on Build GRCh38
rs199836285 Chr.12: 26340239-26340239 on Build GRCh38
rs767675618 Chr.12: 26340276-26340276 on Build GRCh38
rs772677036 Chr.12: 26340218-26340218 on Build GRCh38
rs368577591 Chr.12: 26340138-26340138 on Build GRCh38
rs992421552 Chr.12: 26340279-26340279 on Build GRCh38
rs779028166 Chr.12: 26340199-26340199 on Build GRCh38
rs775898828 Chr.12: 26340358-26340358 on Build GRCh38
rs756462377 Chr.12: 26340142-26340142 on Build GRCh38
rs541786341 Chr.12: 26340183-26340183 on Build GRCh38
rs368039850 Chr.12: 26340283-26340283 on Build GRCh38
rs147676775 Chr.12: 26340351-26340351 on Build GRCh38
rs758494207 Chr.12: 26340164-26340164 on Build GRCh38
rs764229958 Chr.12: 26340265-26340265 on Build GRCh38
rs550210847 Chr.12: 26340229-26340229 on Build GRCh38
rs777312617 Chr.12: 26340318-26340318 on Build GRCh38
rs765682095 Chr.12: 26340242-26340242 on Build GRCh38
rs200692965 Chr.12: 26340195-26340195 on Build GRCh38
rs746537715 Chr.12: 26340320-26340320 on Build GRCh38
rs766774155 Chr.12: 26340151-26340151 on Build GRCh38
rs561869925 Chr.12: 26340187-26340187 on Build GRCh38
rs916537591 Chr.12: 26340214-26340214 on Build GRCh38
rs758337577 Chr.12: 26340310-26340310 on Build GRCh38
rs770147017 Chr.12: 26340352-26340352 on Build GRCh38
rs200691111 Chr.12: 26340222-26340222 on Build GRCh38

View Full Product Details