Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122856258-122856491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRC2
Entrez Gene ID
84925
Gene Name
disrupted in renal carcinoma 2
Gene Aliases
RCC4
UniGene
Hs.477346
Gene Chromosome Location
Chr.3: 122795054-122892157 on Build GRCh38
Transcripts
NM_032839.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309916 c.973A>G p.I325V Chr.3: 122856337-122856337 on Build GRCh38
1037265 c.976G>T p.V326F Chr.3: 122856340-122856340 on Build GRCh38
6261156 c.968G>T p.W323L Chr.3: 122856332-122856332 on Build GRCh38
6327746 c.978T>A p.V326V Chr.3: 122856342-122856342 on Build GRCh38
6488201 c.971C>A p.S324Y Chr.3: 122856335-122856335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546215954 Chr.3: 122856447-122856447 on Build GRCh38
rs760826888 Chr.3: 122856368-122856368 on Build GRCh38
rs577132859 Chr.3: 122856443-122856443 on Build GRCh38
rs780847060 Chr.3: 122856322-122856322 on Build GRCh38
rs930168202 Chr.3: 122856292-122856292 on Build GRCh38
rs1005696578 Chr.3: 122856388-122856388 on Build GRCh38
rs769002109 Chr.3: 122856327-122856327 on Build GRCh38
rs200589785 Chr.3: 122856403-122856403 on Build GRCh38
rs756821005 Chr.3: 122856315-122856315 on Build GRCh38
rs933288251 Chr.3: 122856304-122856304 on Build GRCh38
rs756774828 Chr.3: 122856426-122856426 on Build GRCh38
rs115393196 Chr.3: 122856442-122856442 on Build GRCh38
rs765112895 Chr.3: 122856389-122856389 on Build GRCh38
rs568324197 Chr.3: 122856367-122856367 on Build GRCh38
rs886667270 Chr.3: 122856319-122856319 on Build GRCh38
rs766647865 Chr.3: 122856380-122856380 on Build GRCh38
rs774753636 Chr.3: 122856341-122856341 on Build GRCh38
rs776861161 Chr.3: 122856418-122856418 on Build GRCh38
rs752590221 Chr.3: 122856401-122856401 on Build GRCh38
rs759906822 Chr.3: 122856383-122856383 on Build GRCh38
rs888232825 Chr.3: 122856337-122856337 on Build GRCh38
rs748580026 Chr.3: 122856351-122856351 on Build GRCh38
rs750985479 Chr.3: 122856414-122856414 on Build GRCh38
rs374521927 Chr.3: 122856355-122856355 on Build GRCh38
rs764123952 Chr.3: 122856408-122856408 on Build GRCh38
rs1006454817 Chr.3: 122856342-122856342 on Build GRCh38
rs746569728 Chr.3: 122856307-122856307 on Build GRCh38
rs1012458031 Chr.3: 122856375-122856375 on Build GRCh38

View Full Product Details