Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130911595-130911868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
-
UniGene
Hs.133205
Gene Chromosome Location
Chr.9: 130902438-130939068 on Build GRCh38
Transcripts
NM_001145106.1 NM_032843.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368276 c.907C>T p.R303* Chr.9: 130911831-130911831 on Build GRCh38
3718725 c.768C>G p.Y256* Chr.9: 130911831-130911831 on Build GRCh38
6323295 c.902C>T p.A301V Chr.9: 130911836-130911836 on Build GRCh38
3654931 c.933G>A p.G311G Chr.9: 130911805-130911805 on Build GRCh38
4513049 c.925C>T p.L309F Chr.9: 130911813-130911813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559408176 Chr.9: 130911738-130911738 on Build GRCh38
rs775076933 Chr.9: 130911819-130911819 on Build GRCh38
rs973304218 Chr.9: 130911711-130911711 on Build GRCh38
rs1002996125 Chr.9: 130911616-130911616 on Build GRCh38
rs779520322 Chr.9: 130911842-130911842 on Build GRCh38
rs769347517 Chr.9: 130911817-130911817 on Build GRCh38
rs1012245597 Chr.9: 130911760-130911760 on Build GRCh38
rs538784572 Chr.9: 130911637-130911637 on Build GRCh38
rs1056328583 Chr.9: 130911730-130911730 on Build GRCh38
rs56126622 Chr.9: 130911769-130911769 on Build GRCh38
rs541870250 Chr.9: 130911780-130911780 on Build GRCh38
rs928964313 Chr.9: 130911720-130911720 on Build GRCh38
rs763876802 Chr.9: 130911787-130911787 on Build GRCh38
rs1020793575 Chr.9: 130911743-130911743 on Build GRCh38
rs552451805 Chr.9: 130911847-130911847 on Build GRCh38
rs535024810 Chr.9: 130911665-130911665 on Build GRCh38
rs752104263 Chr.9: 130911745-130911745 on Build GRCh38
rs1049526607 Chr.9: 130911694-130911694 on Build GRCh38
rs925430977 Chr.9: 130911706-130911706 on Build GRCh38
rs769443515 Chr.9: 130911849-130911849 on Build GRCh38
rs184076215 Chr.9: 130911649-130911649 on Build GRCh38
rs768960152 Chr.9: 130911757-130911757 on Build GRCh38
rs377304322 Chr.9: 130911770-130911770 on Build GRCh38
rs550375715 Chr.9: 130911680-130911680 on Build GRCh38
rs750732451 Chr.9: 130911835-130911835 on Build GRCh38
rs918923861 Chr.9: 130911712-130911712 on Build GRCh38
rs770604739 Chr.9: 130911761-130911761 on Build GRCh38
rs565978747 Chr.9: 130911650-130911650 on Build GRCh38
rs550474087 Chr.9: 130911837-130911837 on Build GRCh38
rs570456220 Chr.9: 130911703-130911703 on Build GRCh38
rs762300224 Chr.9: 130911820-130911820 on Build GRCh38
rs750650927 Chr.9: 130911797-130911797 on Build GRCh38
rs199643737 Chr.9: 130911848-130911848 on Build GRCh38
rs756213471 Chr.9: 130911748-130911748 on Build GRCh38
rs896296414 Chr.9: 130911735-130911735 on Build GRCh38
rs368351261 Chr.9: 130911752-130911752 on Build GRCh38
rs780867280 Chr.9: 130911808-130911808 on Build GRCh38
rs768667343 Chr.9: 130911825-130911825 on Build GRCh38
rs368873188 Chr.9: 130911774-130911774 on Build GRCh38
rs753167205 Chr.9: 130911843-130911843 on Build GRCh38
rs1028107757 Chr.9: 130911634-130911634 on Build GRCh38
rs967366218 Chr.9: 130911674-130911674 on Build GRCh38
rs775352223 Chr.9: 130911777-130911777 on Build GRCh38
rs536407048 Chr.9: 130911659-130911660 on Build GRCh38
rs545620826 Chr.9: 130911725-130911725 on Build GRCh38
rs190115223 Chr.9: 130911685-130911685 on Build GRCh38
rs149922280 Chr.9: 130911826-130911826 on Build GRCh38
rs755514768 Chr.9: 130911804-130911804 on Build GRCh38
rs912032057 Chr.9: 130911641-130911641 on Build GRCh38
rs866063692 Chr.9: 130911814-130911814 on Build GRCh38
rs780755370 Chr.9: 130911750-130911750 on Build GRCh38
rs748396074 Chr.9: 130911759-130911759 on Build GRCh38
rs760073718 Chr.9: 130911750-130911752 on Build GRCh38
rs112808686 Chr.9: 130911679-130911679 on Build GRCh38
rs780268824 Chr.9: 130911800-130911800 on Build GRCh38
rs757075901 Chr.9: 130911796-130911796 on Build GRCh38
rs371263618 Chr.9: 130911807-130911807 on Build GRCh38
rs745511635 Chr.9: 130911815-130911815 on Build GRCh38
rs986337271 Chr.9: 130911639-130911639 on Build GRCh38
rs986924724 Chr.9: 130911784-130911784 on Build GRCh38
rs765585062 Chr.9: 130911768-130911769 on Build GRCh38
rs532727695 Chr.9: 130911846-130911846 on Build GRCh38
rs144181457 Chr.9: 130911831-130911831 on Build GRCh38
rs1011804647 Chr.9: 130911656-130911656 on Build GRCh38
rs750656149 Chr.9: 130911805-130911805 on Build GRCh38
rs762931223 Chr.9: 130911781-130911781 on Build GRCh38
rs554494565 Chr.9: 130911677-130911677 on Build GRCh38
rs7044609 Chr.9: 130911709-130911709 on Build GRCh38
rs766443740 Chr.9: 130911836-130911836 on Build GRCh38
rs6597650 Chr.9: 130911838-130911838 on Build GRCh38
rs751371909 Chr.9: 130911792-130911792 on Build GRCh38
rs749270628 Chr.9: 130911801-130911801 on Build GRCh38
rs1017855522 Chr.9: 130911823-130911823 on Build GRCh38

View Full Product Details