Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 104424204-104424465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYS
Entrez Gene ID
1807
Gene Name
dihydropyrimidinase
Gene Aliases
DHP, DHPase
UniGene
Hs.443161
Gene Chromosome Location
Chr.8: 104379424-104467075 on Build GRCh38
Transcripts
NM_001385.2 XM_005250818.3 XM_006716518.3 XM_011516903.2 XM_017013167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
351695 c.1117A>T p.R373* Chr.8: 104424365-104424365 on Build GRCh38
269057 c.1158T>A p.F386L Chr.8: 104424324-104424324 on Build GRCh38
1094643 c.1219G>A p.D407N Chr.8: 104424263-104424263 on Build GRCh38
748886 c.1181G>C p.R394T Chr.8: 104424301-104424301 on Build GRCh38
6111470 c.1136G>T p.S379I Chr.8: 104424346-104424346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774340932 Chr.8: 104424397-104424397 on Build GRCh38
rs267606774 Chr.8: 104424247-104424247 on Build GRCh38
rs201258823 Chr.8: 104424345-104424345 on Build GRCh38
rs565135052 Chr.8: 104424366-104424366 on Build GRCh38
rs367990408 Chr.8: 104424324-104424324 on Build GRCh38
rs76977023 Chr.8: 104424352-104424352 on Build GRCh38
rs745926279 Chr.8: 104424398-104424398 on Build GRCh38
rs763914910 Chr.8: 104424318-104424318 on Build GRCh38
rs760542155 Chr.8: 104424317-104424317 on Build GRCh38
rs755440333 Chr.8: 104424254-104424254 on Build GRCh38
rs374461372 Chr.8: 104424269-104424269 on Build GRCh38
rs781577893 Chr.8: 104424260-104424260 on Build GRCh38
rs747494363 Chr.8: 104424252-104424252 on Build GRCh38
rs1014702313 Chr.8: 104424374-104424374 on Build GRCh38
rs756723920 Chr.8: 104424244-104424244 on Build GRCh38
rs1054181740 Chr.8: 104424257-104424257 on Build GRCh38
rs771149650 Chr.8: 104424389-104424389 on Build GRCh38
rs772200986 Chr.8: 104424408-104424408 on Build GRCh38
rs768357180 Chr.8: 104424413-104424413 on Build GRCh38
rs763824762 Chr.8: 104424241-104424241 on Build GRCh38
rs778144712 Chr.8: 104424382-104424382 on Build GRCh38
rs1028048044 Chr.8: 104424356-104424356 on Build GRCh38
rs141867641 Chr.8: 104424293-104424293 on Build GRCh38
rs749500473 Chr.8: 104424387-104424387 on Build GRCh38
rs764686138 Chr.8: 104424329-104424329 on Build GRCh38
rs372071049 Chr.8: 104424410-104424410 on Build GRCh38
rs147101902 Chr.8: 104424340-104424340 on Build GRCh38
rs749463169 Chr.8: 104424291-104424291 on Build GRCh38
rs770063251 Chr.8: 104424265-104424265 on Build GRCh38
rs759428977 Chr.8: 104424300-104424300 on Build GRCh38
rs775479555 Chr.8: 104424315-104424315 on Build GRCh38
rs753454667 Chr.8: 104424242-104424242 on Build GRCh38
rs781657979 Chr.8: 104424359-104424359 on Build GRCh38
rs776448083 Chr.8: 104424423-104424423 on Build GRCh38
rs1000896221 Chr.8: 104424230-104424230 on Build GRCh38
rs756477018 Chr.8: 104424373-104424373 on Build GRCh38
rs767535896 Chr.8: 104424312-104424312 on Build GRCh38
rs370814653 Chr.8: 104424409-104424409 on Build GRCh38
rs748491306 Chr.8: 104424263-104424263 on Build GRCh38
rs750724321 Chr.8: 104424323-104424323 on Build GRCh38
rs370170070 Chr.8: 104424246-104424246 on Build GRCh38

View Full Product Details